Skip to Content
Merck
All Photos(1)

Key Documents

V3204

Sigma-Aldrich

4-Vinylpyridine

contains 100 ppm hydroquinone as inhibitor, 95%

Synonym(s):

4-Ethenylpyridine Pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N
CAS Number:
Molecular Weight:
105.14
Beilstein:
104506
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

contains

100 ppm hydroquinone as inhibitor

refractive index

n20/D 1.549 (lit.)

bp

62-65 °C/15 mmHg (lit.)

density

0.975 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C=Cc1ccncc1

InChI

1S/C7H7N/c1-2-7-3-5-8-6-4-7/h2-6H,1H2

InChI key

KFDVPJUYSDEJTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Vinylpyridine is a highly reactive alkene monomer due to the presence of a vinyl group and a heteroaromatic ring. It is widely used as a monomer to produce a variety of polymers and copolymers, which are used in various applications including, contact lenses, nanoelectronics, sensors, optoelectronic devices, smart coatings, drug delivery systems, sensors, gas separation, air filtration, and purification processes.

Application

4-Vinylpyridine can be used as a monomer:
  • In the synthesis or modification of polymers for drug-releasing contact lenses.
  • In the preparation of the zwitterionic polymer, namely poly(4-vinylpyridine propylsulfobetaine). The zwitterionic polymer is utilized for its superior bioinert capability in withstanding clinical sterilization processes, making it suitable for extended medical applications which include the development of biocompatible implants and as a coating material for medical devices.
  • To synthesize the temperature and pH-sensitive copolymer, specifically (N-vinylcaprolactam-co-4-vinylpyridine), which is used in drug delivery systems under certain environmental conditions.
  • The highly charged p(4-vinylpyridine-co-vinylimidazole) particles, which find applications in various fields such as biomedical, catalysis, and environmental applications.
4-Vinylpyridine also plays a role as a functional monomer in the molecular imprinting of the soft contact lens material. Its inclusion, along with other monomers, enhances the affinity of the hydrogel towards brimonidine, resulting in improved binding properties, loading capacity, and controlled release characteristics.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.0 °F - closed cup

Flash Point(C)

51.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujuan Zhang et al.
BMC plant biology, 20(1), 45-45 (2020-01-31)
Wheat grain avenin-like proteins (ALPs) belong to a recently discovered class of wheat grain storage protein. ALPs in wheat grains not only have beneficial effects on dough quality but also display antifungal activities, which is a novel observation for wheat
Chadin Kulsing et al.
Electrophoresis, 38(8), 1179-1187 (2017-02-18)
This investigation demonstrates the application of a new peak sharpening technique to improve the separation of difficult-to-resolve racemic mixtures in capillary electro-chromatography. Molecularly imprinted porous layer open tubular (MIP-PLOT) capillaries, prepared by a layer-on-layer polymerization approach with Z-l-Asp-OH as the
Polymer, 34, 145-145 (1993)
Vusumzi Pakade et al.
Journal of chromatography. A, 1230, 15-23 (2012-02-22)
Molecularly imprinted polymers (MIPs) targeting quercetin were prepared from 4-vinylpyridine and ethylene dimethacrylate (EDMA) under various solvent systems with the aim to form MIPs with high recognition for the quercetin molecule in aqueous systems at high temperature. A MIP prepared
Turghun Muhammad et al.
Analytica chimica acta, 709, 98-104 (2011-11-30)
Novel water-compatible molecularly imprinted polymers (MIPs) selective for amiodarone (AD) were designed via a new methodology which relies on screening library of non-imprinted polymers (NIPs). The NIP library consisted of eighteen cross-linked co-polymers synthesized from monomers commonly used in molecular

Articles

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service