Skip to Content
Merck
All Photos(1)

Key Documents

233803

Sigma-Aldrich

3-(Trimethylsilyl)-1-propanol

97%

Synonym(s):

1-(Trimethylsilyl)-3-propanol, 3-(Trimethylsilyl)-1-propanol, 3-(Trimethylsilyl)propanol, 3-(Trimethylsilyl)propyl alcohol, 3-Hydroxypropyltrimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3Si(CH2)3OH
CAS Number:
Molecular Weight:
132.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.428 (lit.)

bp

111-112 °C/96 mmHg (lit.)

density

0.832 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C[Si](C)(C)CCCO

InChI

1S/C6H16OSi/c1-8(2,3)6-4-5-7/h7H,4-6H2,1-3H3

InChI key

YNUUCDHAZGAVEZ-UHFFFAOYSA-N

Application

3-(Trimethylsilyl)-1-propanol has been used in:
  • the synthesis of telechelic oligomers via atom transfer radical polymerization (ATRP) of styrene[1]
  • the preparation of the β-seleno-γ-silyl alcohols[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silicon-controlled rearrangement versus Krief-Reich reaction in ?-seleno-?-silyl alcohols: a new stereoselective synthesis of terminally-substituted allylsilanes.
Sarkar TK and Ghosh SK.
Tetrahedron Letters, 28(18), 2061-2064 (1987)
Synthesis of Telechelic Oligomers via Atom Transfer Radical Polymerization, 1.
Otazaghine B, et al.
Macromolecular Chemistry and Physics, 205(2), 154-164 (2004)
Adam Czajka et al.
Physical chemistry chemical physics : PCCP, 19(35), 23869-23877 (2017-07-21)
Presented here are the results for a novel class of hydrocarbon surfactants, termed trimethylsilyl hedgehogs (TMS-hedgehogs), due to the presence of silicon in the tails. By comparing the surface properties of these hybrid hedgehogs to purely hydrocarbon equivalents, links between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service