Skip to Content
Merck
All Photos(2)

Documents

178837

Sigma-Aldrich

3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt

97%

Synonym(s):

2,2-Dimethyl-2-silapentane-5-sulfonate sodium salt, DSS sodium salt, Sodium 3-(trimethylsilyl)-1-propanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3Si(CH2)3SO3Na
CAS Number:
Molecular Weight:
218.32
Beilstein:
4035923
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

~165 °C (dec.) (lit.)

SMILES string

[Na+].C[Si](C)(C)CCCS([O-])(=O)=O

InChI

1S/C6H16O3SSi.Na/c1-11(2,3)6-4-5-10(7,8)9;/h4-6H2,1-3H3,(H,7,8,9);/q;+1/p-1

InChI key

HWEXKRHYVOGVDA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt can be used as a reagent to synthesize polyamine block-copolymers. It can also be used as an internal standard in 1H NMR spectroscopy for the measurement of chemical shifts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerome Kretzschmar et al.
Inorganic chemistry, 59(7), 4244-4254 (2020-03-10)
The interactions between glutathione disulfide, GSSG, the redox partner and dimer of the intracellular detoxification agent glutathione, GSH, and hexavalent uranium, U(VI), were extensively studied by solution NMR (in D2O), complemented by time-resolved laser-induced fluorescence and IR spectroscopies. As expected
J F Clark et al.
Biochimica et biophysica acta, 1014(3), 235-238 (1989-12-14)
3-(Trimethylsilyl)propanesulfonic acid (TMSPS) is used as a water-soluble NMR frequency marker. It has its major resonance at 0.00 ppm relative to trimethylsilane, and smaller resonances at 0.62, 1.77 and 2.85 ppm. Its toxicity was tested by exposing contracted porcine carotid
Nicole D Wagner et al.
Environmental toxicology and chemistry, 37(9), 2448-2457 (2018-06-20)
Anthropogenic activity is increasing the presence of contaminants that enter waterways through wastewater effluent and urban and/or agricultural runoff, generally in complex mixtures. Depending on the mode of action of the individual contaminant within the mixture, toxicity can occur due
Douglas V Laurents et al.
The Journal of biological chemistry, 280(5), 3675-3685 (2004-11-24)
The Alzheimer's Abeta40 peptide forms soluble oligomers that are extremely potent neurotoxins and strongly impede synapses function. In this study the formation and structure of the large, soluble, neurotoxic Abeta40 oligomer called "beta-ball" were characterized by two-dimensional NMR, circular dichroism
Dong Min Han et al.
Food microbiology, 92, 103591-103591 (2020-09-21)
Six different ganjang batches were prepared and the microbial communities in the ganjang samples and raw materials (meju and solar salts), as well as the metabolites generated during fermentation were analyzed. The varying amounts of raw materials used differentially affected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service