Skip to Content
Merck
All Photos(1)

Key Documents

233412

Sigma-Aldrich

Ethyl (hydroxyimino)cyanoacetate

97%, for peptide synthesis

Synonym(s):

Ethyl cyano(hydroxyimino)acetate, Ethyl cyanoglyoxalate-2-oxime, Ethyl isonitrosocyanoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC(=NOH)CO2C2H5
CAS Number:
Molecular Weight:
142.11
Beilstein:
774783
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product name

Ethyl (hydroxyimino)cyanoacetate, 97%

Assay

97%

form

solid

mp

130-132 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCOC(=O)C(=N\O)\C#N

InChI

1S/C5H6N2O3/c1-2-10-5(8)4(3-6)7-9/h9H,2H2,1H3/b7-4+

InChI key

LCFXLZAXGXOXAP-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl (hydroxyimino)cyanoacetate is a potential replacement for zobenzotriazole and benzotriazole derivatives used in peptide synthesis.

Ethyl (hydroxyimino)cyanoacetate is also called as Oxyma. It is a highly efficient greener alternative for the amide and peptide synthesis.

Application

Ethyl (hydroxyimino)cyanoacetate has been used as an additive for the carbodiimide-mediated amide bond formation during established peptide synthesis method.For peptide synthesis grade material, please see product 851086.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minimizing HCN in DIC/Oxyma-mediated amide bond-forming reactions
Erny, Marion and Lundqvist, et al.
Organic Process Research & Development, 24, 1341-1349 (2020)
Ranko Gacesa et al.
International journal of molecular sciences, 21(20) (2020-10-16)
Siderophores are iron-complexing compounds synthesized by bacteria and fungi. They are low molecular weight compounds (500-1500 Daltons) possessing high affinity for iron(III). Since 1970 a large number of siderophores have been characterized, the majority using hydroxamate or catecholate as functional
Miriam Reverter et al.
Metabolites, 10(6) (2020-06-04)
Understanding natural defense mechanisms against parasites can be a valuable tool for the development of innovative therapies. We have previously identified a butterflyfish species (Chaetodonlunulatus) that avoids gill monogenean parasites while living amongst closely related parasitized species. The metabolome and
Daniele Maiolo et al.
ChemistryOpen, 9(2), 253-260 (2020-02-29)
Here, we demonstrate that introduction of halogen atoms at the tyrosine 10 phenol ring of the DSGYEV sequence derived from the flexible amyloid-β N-terminus, promotes its self-assembly in the solid state. In particular, we report the crystal structures of two halogen-modified
Hangyu Zhang et al.
Acta biomaterialia, 55, 183-193 (2017-04-04)
Self-assembling peptides programed by sequence design to form predefined nanostructures are useful for a variety of biomedical applications. However, assemblies of classic ionic self-complementary peptides are unstable in neutral pH, while charged peptide hydrogels have low mechanical strength. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service