Skip to Content
Merck
All Photos(1)

Key Documents

56413

Sigma-Aldrich

2-Pyridinol 1-oxide

≥98.0% (N)

Synonym(s):

1-Hydroxy-2-pyridone, 2-Hydroxypyridine 1-oxide, HOPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO2
CAS Number:
Molecular Weight:
111.10
Beilstein:
1422545
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (N)

form

solid

mp

147-152 °C

SMILES string

Oc1cccc[n+]1[O-]

InChI

1S/C5H5NO2/c7-5-3-1-2-4-6(5)8/h1-4,7H

InChI key

JVHZMYAXZUIZKS-UHFFFAOYSA-N

General description

2-Pyridinol 1-oxide (HOPO) is a peptide coupling agent used as a subsititue for HOBt. It can form stable chelate complexes with transition metal ions.

Application

2-Pyridinol 1-oxide can be used:    
  • To fabricate gold electrodes for the electrochemical determination of europium using cyclic voltammetry.        
  • To prepare polymeric nickel (II) complex of 2-hydroxypyridine-N-oxide.
  • In the synthesis of 1-methoxypyrid-4-one in the presence of sodium methoxide and methyl toluene-9-sulphonate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Cruickshank et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(7), 623-627 (2015-07-15)
This work presents for the first time the electrochemical determination of europium using cyclic voltammetry at gold electrodes modified with 2-pyridinol-1-oxide. A well-defined oxidation peak was observed in cyclic voltammetry as a result of the oxidation of the europium at
Structure and Behavior of Organic Analytical Reagents. Formation Constants of Transition Metal Complexes of 2-Hydroxypyridine-1-Oxide and 2-Mercaptopyridine-1-Oxide.
Sun PJ, et al.
Analytical Chemistry, 36(13), 2485-2488 (1964)
Dong-Geon Lee et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 20(5), 752-756 (2019-01-11)
Porous carbonaceous anode materials have received considerable attention as an alternative anode material, however, there is a critical bottleneck as it suffers from a large irreversible specific capacity loss over several initial cycles owing to undesired surface reactions. In order
875. N-oxides and related compounds. Part V. The tautomerism of 2-and 4-amino-and-hydroxy-pyridine 1-oxide.
Gardne JN & Katritzky AR.
Journal of the Chemical Society, 4375-4385 (1957)
Ali Younes et al.
Metallomics : integrated biometal science, 11(2), 496-507 (2019-01-16)
Uranium is widespread in the environment, resulting both from natural occurrences and anthropogenic activities. Its toxicity is mainly chemical rather than radiological. In the blood it is transported as uranyl UO22+ cation and forms complexes with small ligands like carbonates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service