Skip to Content
Merck
All Photos(1)

Key Documents

38370

Sigma-Aldrich

DIC

≥98.0% (GC), for peptide synthesis

Synonym(s):

N,N′-Diisopropylcarbodiimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHN=C=NCH(CH3)2
CAS Number:
Molecular Weight:
126.20
Beilstein:
878281
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

product name

DIC, purum, ≥98.0% (GC)

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

liquid

reaction suitability

reaction type: Coupling Reactions

refractive index

n20/D 1.433 (lit.)

bp

145-148 °C (lit.)

density

0.815 g/mL at 20 °C (lit.)
0.815 g/mL at 20 °C

application(s)

peptide synthesis

SMILES string

CC(C)N=C=NC(C)C

InChI

1S/C7H14N2/c1-6(2)8-5-9-7(3)4/h6-7H,1-4H3

InChI key

BDNKZNFMNDZQMI-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Diisopropylcarbodiimide (DIC) is a carbodiimide used as a coupling reagent in the synthesis of amides, peptides, ureas, heterocycles, and unsymmetrical carbodiimides. It is also used in the polymerization reactions as an activator.

Application

N,N′-Diisopropylcarbodiimide can be used:
  • To synthesize lanthanide (Ln) guanidinate complexes via insertion of carbodiimide into the Ln-N bond of lanthanocene secondary amido complexes.
  • To facilitate the cyclization of N-(β-Hydroxy)amides to form 2-oxazolines.
  • To synthesize 1-isopropyl-2-alkoxycarbonyl-3-isopropyliminio-aziridine by reacting with alkyl diazoacetates in the presence of transition metal salts.

DIC can be used as:
  • A coupling reagent for the synthesis of various esters and amides by treating carboxylic acids with phenols and amines respectively.
  • A reagent for the conversion of alcohols to aldehydes or ketones in the presence of DMSO via modified Moffatt-type oxidation reaction.
  • A reagent to facilitates the preparation of alkyl halides from corresponding alcohols via the formation of o-alkylisourea.

Alternative to dicyclohexylcarbodiimide in peptide synthesis.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F

Flash Point(C)

33 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 3-Diisopropylcarbodiimide
Nora GP, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Insertion of a carbodiimide into the Ln? N ?-bond of organolanthanide complexes. Isomerization and rearrangement of organolanthanides containing guanidinate ligands.
Zhang J
Organometallics, 23(13), 3303-3308 (2004)
Amidation and esterification of carboxylic acids with amines and phenols by N, N?-diisopropylcarbodiimide: A new approach for amide and ester bond formation in water
Fattahi N, et al.
Tetrahedron, 74(32), 4351-4356 (2018)
Stefano Crosignani et al.
Chemical communications (Cambridge, England), (2), 260-261 (2003-02-15)
Alcohols can be converted in high yields to the corresponding alkyl halides in a one-pot procedure via the corresponding O-alkylisourea; very short reaction times are possible when microwave irradiation is used.
Synthesis of iminoaziridines from carbodiimides and diazoesters: a new example of transition metal salt catalysed reactions of carbenes.
Hubert A
Tetrahedron Letters, 17(16), 1317-1318 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service