Skip to Content
Merck
All Photos(3)

Key Documents

200654

Sigma-Aldrich

Ethyl cellulose

viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl

Synonym(s):

Ethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

granular

Quality Level

autoignition temp.

698 °F

concentration

48.0-49.5%

extent of labeling

48% ethoxyl

refractive index

n20/D 1.47 (lit.)

viscosity

300 cP, 5 % in toluene/ethanol 80:20(lit.)

transition temp

softening point 157 °C

density

1.14 g/mL at 25 °C (lit.)

SMILES string

O1[C@H](C(C(C(C1COCC)OCC)OCC)OCC)O[C@@H]2C(OC(C(C2OCC)OCC)OCC)COCC

InChI

1S/C28H54O11/c1-9-29-17-19-21(31-11-3)23(32-12-4)26(35-15-7)28(38-19)39-22-20(18-30-10-2)37-27(36-16-8)25(34-14-6)24(22)33-13-5/h19-28H,9-18H2,1-8H3/t19?,20?,21?,22-,23?,24?,25?,26?,27?,28+/m1/s1

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl cellulose (EC) is a non-toxic, biocompatible, and hydrophobic polymer used for the sustained release preparation of pharmaceutical products.

Application

Ethyl cellulose can be used to prepare polymer coatings for various applications. For example, it is used to encapsulate salt hydrates for low-temperature heat storage applications.

It can be used as an efficient carrier for multiparticulate sustained drug delivery. For example, it can be used to develop polymeric microspheres for the controlled release of Ivabradine HCL (IBH).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dace Gao et al.
Nanoscale horizons, 5(4), 730-738 (2020-02-18)
The design and construction of 3D architectures enabled by stimuli-responsive soft materials can yield novel functionalities for next generation soft-bodied actuating devices. Apart from additive manufacturing processes, origami inspired technology offers an alternative approach to fabricate 3D actuators from planar
Kifayat Ullah Shah et al.
TheScientificWorldJournal, 2012, 842348-842348 (2012-06-01)
The design and fabrication of sustained/controlled release dosage forms, employing new excipients capable of extending/controlling the release of drugs from the dosage forms over prolonged periods, has worked well in achieving optimally enhanced therapeutic levels of the drugs. In this
Ghulam Murtaza
Acta poloniae pharmaceutica, 69(1), 11-22 (2012-05-12)
Ethylcellulose (EC) based microencapsulated drug delivery systems are being extensively studied throughout the world for achieving extended drug release and protecting the core substance from degradation. The in vitro evaluation of EC microcapsules have elucidated that their particle characteristics are
Oguzhan Gunduz et al.
Pharmaceutical research, 30(1), 225-237 (2012-09-08)
To investigate a new microfluidic method for the continuous preparation of hollow-shell nanoparticles of a hydrophobic polymer and the simultaneous encapsulation within these of a hydrophilic active pharmaceutical ingredient. A specially designed and constructed microfluidic device which facilitates at a
Ritesh M Pabari et al.
Expert opinion on drug delivery, 9(12), 1463-1474 (2012-10-16)
To examine the potential of a novel 3-fluid nozzle spray drying technology to formulate differentiated layered microparticles (MPs) of diclofenac sodium (DFS)/ethyl cellulose (EC). DFS/EC MPs were formulated using the inner and/or outer nozzles of a novel 3-fluid nozzle and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service