Skip to Content
Merck
All Photos(4)

Key Documents

185329

Sigma-Aldrich

3-Nitrobenzoic acid

ReagentPlus®, 99%

Synonym(s):

m-Carboxynitrobenzene, m-Nitrobenzenecarboxylic acid, m-Nitrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CO2H
CAS Number:
Molecular Weight:
167.12
Beilstein:
908644
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39032047
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

Assay

99%

form

solid

mp

139-141 °C (lit.)

solubility

water: soluble 3 g/L at 25 °C

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1cccc(c1)[N+]([O-])=O

InChI

1S/C7H5NO4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H,(H,9,10)

InChI key

AFPHTEQTJZKQAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Nitrobenzoic acid forms molecular adducts with commercially important herbicides and pesticides such as amitrole.

Application

3-Nitrobenzoic acid was used to investgate the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

373.3 °F - Pensky-Martens closed cup

Flash Point(C)

189.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular co-crystals of carboxylic acids. 16. 1: 1 Adduct of 3-nitrobenzoic acid with 3-amino-1H-1, 2, 4-triazole.
Lynch DE, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 50(8), 1291-1294 (1994)
W Jiang et al.
The Analyst, 122(3), 211-215 (1997-03-01)
In an attempt to produce complete oxidation of a biological matrix, bovine liver, ozone was investigated as an additional, potentially non-contaminating, oxidizing reagent after nitric acid digestion. Experiments were carried out to determine the decomposition efficiency of residual carbon species
M Algarra et al.
Talanta, 83(5), 1335-1340 (2011-01-18)
The detection of nitroaromatic compounds, best known as raw materials in explosives preparations, is important in many fields including environmental science, public security and forensics. CdSe quantum dots capped with PAMAM-G(4) dendrimer were synthetized in water and used for the
B D'eng et al.
Voprosy meditsinskoi khimii, 29(6), 113-117 (1983-11-01)
Hepatotoxic effects of 4-chloro-3-nitrobenzoic acid (x-NBA), 3-nitrobenzoic acid (NBA) and 4-chlorobenzoic acid (CBA) were studied at the doses corresponding to LD50, 1/10 LD50 and 1/50 LD50. The toxic effects were estimated by monitoring alterations in activity of protein-synthesizing system in
Reproductive toxicology. m-Nitrobenzoic acid.
Environmental health perspectives, 105 Suppl 1, 325-326 (1997-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service