Skip to Content
Merck
All Photos(1)

Documents

137677

Sigma-Aldrich

2-Bromobenzoic acid

97%

Synonym(s):

o-Bromobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CO2H
CAS Number:
Molecular Weight:
201.02
Beilstein:
971266
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

>1 mmHg ( 20 °C)

Assay

97%

form

powder

mp

147-150 °C (lit.)

solubility

95% ethanol: soluble 100 mg/mL, clear, colorless to yellow

SMILES string

OC(=O)c1ccccc1Br

InChI

1S/C7H5BrO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)

InChI key

XRXMNWGCKISMOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Bromobenzoic acid (2-BA) on condensation with aminoquinolines yields phenylquinolylamines. 2-BA is the common building block for synthesis of various nitrogen heterocycles.

Application

2-Bromobenzoic acid was used in the synthesis of new zinc(II) − 2-bromobenzoate complex compounds. It was used as the starting reagent in the synthesis of hemilabile benzimidazolyl phosphine ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition and antimicrobial activity of zinc (II) 2-bromobenzoates with organic ligands.
Krajnikova A, et al.
Journal of Thermal Analysis and Calorimetry, 105(2), 451-460 (2011)
J B Bongui et al.
Chemical & pharmaceutical bulletin, 49(9), 1077-1080 (2001-09-18)
Condensation of either 2-bromobenzoic acid (4) or 2-chloro-3-nitrobenzoic acid (5) with suitable aminoquinolines 6-8 afforded phenylquinolylamines 9-13. Acid mediated cyclization gave the corresponding 12H-benzo[b][1,7]phenanthrolin-7-ones 14 and 15, and 12H-benzo[b][1,10]phenanthrolin-7-ones 16-18. Compounds 14, 16, and 17 were subsequently N-methylated to 6-demethoxyacronycine
Free radical reactions for heterocycle synthesis. Part 6: 2-Bromobenzoic acids as building blocks in the construction of nitrogen heterocycles.
Zhang W and Pugh G.
Tetrahedron, 59(17), 3009-3018 (2003)
Kin Ho Chung et al.
Chemical communications (Cambridge, England), 48(14), 1967-1969 (2012-01-12)
A new class of easily accessible hemilabile benzimidazolyl phosphine ligands has been developed. The ligand skeleton is prepared from commercially available and inexpensive o-phenylenediamine and 2-bromobenzoic acid. With catalyst loading down to 0.5 mol% palladium, excellent catalytic activity towards the
K H Engesser et al.
FEMS microbiology letters, 51(1), 143-147 (1989-07-15)
Pseudomonas putida strain CLB 250 (DSM 5232) utilized 2-bromo-, 2-chloro- and 2-fluorobenzoate as sole source of carbon and energy. Degradation is suggested to be initiated by a dioxygenase liberating halide in the first catabolic step. After decarboxylation and rearomatization catechol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service