Skip to Content
Merck
All Photos(2)

Key Documents

171883

Sigma-Aldrich

1-Iodopropane

99%

Synonym(s):

Propyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2I
CAS Number:
Molecular Weight:
169.99
Beilstein:
505937
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

43 mmHg ( 25 °C)

Quality Level

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.504 (lit.)

bp

101-102 °C (lit.)

mp

−101 °C (lit.)

density

1.743 g/mL at 25 °C (lit.)

functional group

alkyl halide
iodo

SMILES string

CCCI

InChI

1S/C3H7I/c1-2-3-4/h2-3H2,1H3

InChI key

PVWOIHVRPOBWPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Iodopropane can be used for the alkylation of pyrrole by reacting with potassium salts of pyrrole to synthesize 1-n-propylpyrrole. It can also be used as an n-alkyl iodide model for the vibrational studies.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qun He et al.
Yi chuan = Hereditas, 26(3), 361-363 (2005-01-11)
Suggested a new blocking method for DNA microarray: 1-iodo-propane. We compared the difference between the two block reagents and the blocking results in different blocking time. The results showed that 1-I-propane is batter-blocking reagent than succinic anhydride in DNA chip.
Laura K G Youngster et al.
Applied microbiology and biotechnology, 80(6), 1113-1120 (2008-09-18)
Methyl tert-butyl ether (MTBE) contamination is widespread in aquifers near urban areas around the world. Since this synthetic fuel oxygenate is resistant to most physical methods of treating fuel-contaminated water, biodegradation may be a useful means of remediation. Currently, information
Xiaocun Yang et al.
Bioengineered, 8(1), 14-20 (2016-10-30)
The objective is to explore the effect of testosterone on the proliferation and collagen synthesis of neonatal rat cardiac fibroblasts (CF) induced by Angiotensin II (Ang II) and the underlying mechanisms. Derived from neonatal rats, the CFs were divided into
Jim Moselhy et al.
International journal of nanomedicine, 4, 153-164 (2009-09-24)
Thermoresponsive cationic nanogel (TCNG) networks based on N-isopropylacrylamide (NIPAM), 2-(dimethylamino)ethyl methacrylate (DMAEMA), and quaternary alkyl ammonium halide salts of DMAEMA (DMAEMAQ) were synthesized by dispersion polymerization technique. The thermoresponsive properties of TCNGs and TCNG-salmon sperm DNA (sasDNA) polyplexes were characterized
Stina Syvänen et al.
BMC medical imaging, 7, 6-6 (2007-08-01)
The previously validated NK1-receptor ligand [O-methyl-11C]GR205171 binds with a high affinity to the NK1-receptor and displays a slow dissociation from the receptor. Hence, it cannot be used in vivo for detecting concentration changes in substance P, the endogenous ligand for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service