Skip to Content
Merck
All Photos(3)

Key Documents

167304

Sigma-Aldrich

1-Iodobutane

99%, contains copper as stabilizer

Synonym(s):

Butyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3I
CAS Number:
Molecular Weight:
184.02
Beilstein:
1420755
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

~5 (vs air)

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.498 (lit.)

bp

130-131 °C (lit.)

mp

−103 °C (lit.)

solubility

alcohol: soluble
diethyl ether: soluble
water: insoluble

density

1.617 g/mL at 25 °C (lit.)

functional group

alkyl halide
iodo

SMILES string

CCCCI

InChI

1S/C4H9I/c1-2-3-4-5/h2-4H2,1H3

InChI key

KMGBZBJJOKUPIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thermal chemistry of 1-iodobutane has been investigated on clean and hydrogen- and deuterium-predosed Pt(111) single-crystal surfaces by temperature-programmed desorption and reflection-absorption infrared spectroscopy.

1-Iodobutane is an alkyl halide and is used as an electrophile during SN2 nucleophilic reaction.

Application

1-Iodobutane was used in the synthesis of:
  • S-alkylated 5-(2-,3- and 4-methoxyphenyl)-4H-1,2,4-triazole-3-thiol
  • 5-(2-,3- and 4-methoxyphenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol
  • N-butyl-N-methylpyrrolidinium bis(trifluoromethanesulfonyl)imide by reaction with 1-methylpyrrolidine in ethyl acetate

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Labanauskas et al.
Farmaco (Societa chimica italiana : 1989), 59(4), 255-259 (2004-04-15)
New S-alkylated 5-(2-,3- and 4-methoxyphenyl)-4H-1,2,4-triazole-3-thiols (5a-c, 6a-c) and 5-(2-,3- and 4-methoxyphenyl)-4-phenyl-4H-1,2,4-triazole-3-thiols (7a-c, 8a-c, 9a-c) were synthesized by the alkylation of 3-(2-,3- and 4-methoxyphenyl)-4,5-dihydro-1H-1,2,4-triazole-5-thiones (3a-c) or 3-(2-,3- and 4-methoxyphenyl)-4-phenyl-4,5-dihydro-1H-1,2,4-triazole-5-thiones (4a-c) with 1-iodobutane or 1-(1,3-benzodioxol-5-yl)-2-bromo-1-ethanone, 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)-1-ethanone and 2-bromo-1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)-1-ethanone. Compounds 3a-c and 4a-c
Ilkeun Lee et al.
The journal of physical chemistry. B, 109(7), 2745-2753 (2006-07-21)
The thermal chemistry of a number of C4 hydrocarbons (1,3-butadiene, 1-bromo-3-butene, 1-bromo-2-butene, trans-2-butene, cis-2-butene, 1-butene, 2-iodobutane, 1-iodobutane, and butane) was investigated on clean and hydrogen- and deuterium-predosed Pt(111) single-crystal surfaces by temperature-programmed desorption and reflection-absorption infrared spectroscopy. A combination of
Y Zea-Ponce et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 45(1), 63-68 (1994-01-01)
The benzodiazepine receptor tracer [123I]iomazenil (Ro 16-0154, IMZ) can be prepared in close to theoretical specific activity by the reaction of its tributyltin precursor (IMZ-SnBu3) with [123I]NaI in the presence of Iodogen. However, the labeling reaction is associated with variably
Recent developments in the ENEA lithium metal battery project.
Shin J-H, et al.
Electrochimica Acta, 50(19), 3859-3865 (2005)
NMR Kinetics of the SN2 Reaction between BuBr and I--: An Introductory Organic Chemistry Laboratory Exercise
Mobley, T Andrew
Journal of Chemical Education, 92, 534-537 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service