Skip to Content
Merck
All Photos(1)

Key Documents

8.22290

Sigma-Aldrich

2-Naphthol

for synthesis

Synonym(s):

2-Naphthol, 2-Hydroxy naphthaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O
CAS Number:
Molecular Weight:
144.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
205-182-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.1 hPa ( 30 °C)

Quality Level

Assay

≥99% (GC)

form

solid

autoignition temp.

430 °C

potency

1960 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

bp

294.8 °C/1013 hPa

mp

121.6 °C

transition temp

flash point 153 °C

solubility

1 g/L

density

1.27 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

2-30°C

SMILES string

Oc1cc2c(cc1)cccc2

InChI

1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H

InChI key

JWAZRIHNYRIHIV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 119 °C
Melting range (upper value): ≤ 122 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service