Skip to Content
Merck
All Photos(1)

Key Documents

136123

Sigma-Aldrich

trans-Chalcone

97%

Synonym(s):

Benzylideneacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOC6H5
CAS Number:
Molecular Weight:
208.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

bp

208 °C/25 mmHg (lit.)

mp

55-57 °C (lit.)

functional group

ketone
phenyl

SMILES string

[H]\C(=C(\[H])C(=O)c1ccccc1)c2ccccc2

InChI

1S/C15H12O/c16-15(14-9-5-2-6-10-14)12-11-13-7-3-1-4-8-13/h1-12H/b12-11+

InChI key

DQFBYFPFKXHELB-VAWYXSNFSA-N

Gene Information

human ... IL1B(3553)
rat ... Ar(24208)

Related Categories

General description

trans-Chalcone is an open chain flavonoid that may prevent lung and forestomach cancer.[1]

Application

trans-Chalcone was used in the synthesis of cis and trans diphenyl cyclopropane[2]. It was also used in screening of surface adsorbed species of isobutybenzene and 4-isobutylacetophenone on bulk fosfotungstic Wells-Dawson acid (H6P2W18O62.xH2O)[3].

Biochem/physiol Actions

trans-Chalcone exhibits antifungal activity against Trichophyton rubrum. It is inhibitor of fatty acid synthase[4] and α-amylase[5]. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots[6].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Kun Chiang et al.
Journal of medicinal chemistry, 52(14), 4221-4233 (2009-06-11)
A pharmacophore model, Hypo1, was built on the basis of 21 training-set indole compounds with varying levels of antiproliferative activity. Hypo1 possessed important chemical features required for the inhibitors and demonstrated good predictive ability for biological activity, with high correlation
Saeed Attar et al.
Bioorganic & medicinal chemistry, 19(6), 2055-2073 (2011-02-26)
A series of 30 organic chlacones and 33 ferrocenyl (Fc) chalcones were synthesized and characterized by melting point, elemental analysis, spectroscopy ((1)H NMR and FTIR) and, in two cases, by X-ray crystallography. The biological activity of each compound (10(-4)M in
Asha Budakoti et al.
European journal of medicinal chemistry, 44(3), 1317-1325 (2008-04-02)
In an effort to develop potent antiamoebic agents, we have synthesized chalcones (1-8), amino-5-substituted-(3-phenyl(2-pyrazolinyl))methane-1-thione derivatives (1a-8a) and 2-(5-substituted-3-phenyl-2-pyrazolinyl)-1,3-thiazolino[5,4-b]quinoxaline derivatives (1b-8b) and evaluated for their in vitro antiamoebic activity against HM1:IMSS strain of E. histolytica. All the compounds were characterized by
Wattenberg L.W., et al.
Cancer Letters, 15, 165-165 (1994)
Sylvie Ducki et al.
Bioorganic & medicinal chemistry, 17(22), 7698-7710 (2009-10-20)
The alpha-methyl chalcone SD400 is a potent inhibitor of tubulin assembly and possesses potent anticancer activity. Various chalcone analogues were synthesized and evaluated for their cell growth inhibitory properties against the K562 human chronic myelogenous leukemia cell line (SD400, IC(50)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service