Skip to Content
Merck
All Photos(2)

Key Documents

D204609

Sigma-Aldrich

1,3-Diphenyl-2-propanone

99%

Synonym(s):

1,3-Diphenylacetone, Dibenzyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2CO
CAS Number:
Molecular Weight:
210.27
Beilstein:
974767
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

330 °C (lit.)

mp

32-34 °C (lit.)

SMILES string

O=C(Cc1ccccc1)Cc2ccccc2

InChI

1S/C15H14O/c16-15(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

YFKBXYGUSOXJGS-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Diphenyl-2-propanone is used as a precursor to synthesize structurally defined, fluorescent polyphenylene dendrimers as light emitters for organic light emitting diodes. It is also used in the synthesis of several other polycyclic aromatic hydrocarbons and conjugated polymers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structurally-defined, sulfo-phenylated, oligophenylenes and polyphenylenes.
Skalski T J, et al.
Journal of the American Chemical Society, 137(38), 12223-12226 (2015)
Triphenylene-based polymers for blue polymeric light emitting diodes.
Saleh M, et al.
Macromolecules, 43(1), 137-143 (2009)
Fluoranthene derivatives as blue fluorescent materials for non-doped organic light-emitting diodes.
Kumar S, et al.
J. Mater. Sci., 4(1), 193-200 (2016)
Polytriphenylene Dendrimers: A Unique Design for Blue?Light?Emitting Materials.
Qin T, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 120(43), 8416-8420 (2008)
New hyperbranched conjugated polymers containing hexaphenylbenzene and oxadiazole units: convenient synthesis and efficient deep blue emitters for PLEDs application.
Li Z A, et al.
The Journal of Physical Chemistry B, 114(28), 9101-9108 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service