Skip to Content
Merck
All Photos(4)

Key Documents

127671

Sigma-Aldrich

3-Aminobenzoic acid

98%, for peptide synthesis

Synonym(s):

3-Aminobenzenecarboxylic acid, 3-Carboxyaniline, m-Aminobenzoic acid, Aniline-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
Beilstein:
471603
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Product Name

3-Aminobenzoic acid, 98%

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

178-180 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cccc(c1)C(O)=O

InChI

1S/C7H7NO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

XFDUHJPVQKIXHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lari Lehtiö et al.
Journal of medicinal chemistry, 52(9), 3108-3111 (2009-04-10)
Poly(ADP-ribose) polymerases (PARPs) activate DNA repair mechanisms upon stress- and cytotoxin-induced DNA damage, and inhibition of PARP activity is a lead in cancer drug therapy. We present a structural and functional analysis of the PARP domain of human PARP-3 in
A Yamamoto et al.
Journal of pharmaceutical sciences, 80(11), 1067-1071 (1991-11-01)
Absorption and metabolic characteristics of p-aminobenzoic acid (PABA) and m-aminobenzoic acid (MABA) from the rat small intestine were examined by means of in situ recirculation and in vitro everted sac experiments. p-Aminobenzoic acid was extremely rapidly absorbed from the rat
Seok Min Yoon et al.
Langmuir : the ACS journal of surfaces and colloids, 23(23), 11875-11882 (2007-10-09)
One-dimensional (1D) helical organic nanostructures were synthesized by a modified vapor-solid (VS) process, called the vaporization-condensation-recrystallization (VCR) process. The conventional solution-phase synthetic methods generally mediate self-assemblies of repeating unit molecules. To provide enough intermolecular interaction forces among the unit molecules
Satoru Kamoda et al.
Journal of chromatography. A, 1050(2), 211-216 (2004-10-29)
Carbohydrate chains in glycoprotein pharmaceuticals have important roles for the expression of their biological activities. Therefore, development of an assessment method for the carbohydrate chains is an important parameter for quality control of glycoprotein pharmaceuticals such as newly developed therapeutic
M Nagao et al.
Environmental health perspectives, 93, 169-174 (1991-06-01)
A poly(ADP-ribose) polymerase inhibitor, benzamide (BA), was found to induce flat revertants of NIH 3T3 cells that had been transformed by human Ha-ras, rat Ki-ras, rat c-raf, and human ret-II. These genes had been amplified in original transformants, but they

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service