Skip to Content
Merck
All Photos(3)

Key Documents

D0787

Sigma-Aldrich

3-(Dimethylamino)benzoic acid

≥97% (HPLC), powder

Synonym(s):

3-Dimethylaminobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CO2H
CAS Number:
Molecular Weight:
165.19
Beilstein:
2208586
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

3-(Dimethylamino)benzoic acid, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

mp

148-150 °C (lit.)

solubility

methanol: 50 mg/mL, clear to very slightly hazy, yellow to brown

SMILES string

CN(C)c1cccc(c1)C(O)=O

InChI

1S/C9H11NO2/c1-10(2)8-5-3-4-7(6-8)9(11)12/h3-6H,1-2H3,(H,11,12)

InChI key

NEGFNJRAUMCZMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3-(Dimethylamino)benzoic acid has been used to analyse glucose content from the extraction of starch and soluble sugars.

Biochem/physiol Actions

3-dimethylaminobenzoic acid (DMAB) can be used to quantity the activity of peroxidase and manganese peroxidase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunhua Ding et al.
Journal of integrative plant biology, 58(1), 29-47 (2015-06-03)
Glutathione reductase (GR) catalyzes the reduction of glutathione disulfide (GSSG) to reduced glutathione (GSH) and participates in the ascorbate-glutathione cycle, which scavenges H2 O2 . Here, we report that chloroplastic/mitochondrial GR2 is an important regulator of leaf senescence. Seed development
Temporal and light-based changes in carbon uptake and storage in the spring ephemeral Podophyllum peltatum (Berberidaceae)
Constable J V H, et al.
Environmental and Experimental Botany, 60(1), 112-120 (2007)
M P Castillo et al.
Analytical biochemistry, 218(2), 399-404 (1994-05-01)
This method was proposed earlier for measuring glucose in a peroxidase-glucose oxidase system but has not been studied for determination of manganese peroxidase (MnP) activity. The assay is based on the oxidative coupling of 3-methyl-2-benzothiazolinone hydrazone (MBTH) and 3-(dimethylamino)benzoic acid
Jun Zhao et al.
Plant physiology, 182(1), 640-657 (2019-11-02)
Histone H2B monoubiquitination (H2Bub1) plays critical roles in regulating growth and development as well as stress responses in Arabidopsis (Arabidopsis thaliana). In this study, we used wild-type and HUB1 and HUB2 loss-of-function Arabidopsis plants to elucidate the mechanisms involved in
Potential interference of copper in estimations using Ngo-Lenhoff system.
W L Baker et al.
Analytical biochemistry, 219(2), 386-388 (1994-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service