Skip to Content
Merck
All Photos(2)

Key Documents

126292

Sigma-Aldrich

3,5-Dimethoxybenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CHO
CAS Number:
Molecular Weight:
166.17
Beilstein:
2042374
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

151 °C/16 mmHg (lit.)

mp

45-48 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1cc(OC)cc(OC)c1

InChI

1S/C9H10O3/c1-11-8-3-7(6-10)4-9(5-8)12-2/h3-6H,1-2H3

InChI key

VFZRZRDOXPRTSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used frequently as synthetic building block.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>233.6 °F

Flash Point(C)

> 112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirsti Parikka et al.
Beilstein journal of organic chemistry, 5, 22-22 (2009-07-11)
The first synthesis of bioactive long alkyl chain 5-n-alkylresorcinols, present in whole grain products, by a novel modification of the Wittig reaction is described. All the main long chain 5-n-alkylresorcinols present in rye and wheat, including C(23) and C(25) analogues
Johannes Taeger et al.
Oncology letters, 16(1), 654-659 (2018-06-22)
Despite partial advances in therapy for patients suffering from head and neck squamous cell carcinomas (HNSCC), prognosis still remains poor with minimal improvement in survival for over the last several decades. Some agents found are known to cause cancer cell
Katie A Keaton et al.
Organic letters, 9(14), 2717-2719 (2007-06-15)
A strategy for ketone synthesis with cyclopropanols as intermediates and its application to (+)-spirolaxine methyl ether is described. The synthesis also features an application of Fu's alkyl-alkyl Suzuki coupling.
Monika Osińska-Jaroszuk et al.
Bioprocess and biosystems engineering, 41(7), 973-989 (2018-03-28)
Four bacterial EPSs extracted from Rhizobium leguminosarum bv. trifolii Rt24.2, Sinorhizobium meliloti Rm1021, Bradyrhizobium japonicum USDA110, and Bradyrhizobium elkanii USDA76 were determined towards their metal ion adsorption properties and possible modification of Cerrena unicolor laccase properties. The highest magnesium and
Marco Bandini et al.
Organic letters, 9(11), 2151-2153 (2007-05-08)
A new poly(ethylene glycol)-modified DAT2-Cu(OAc)2 complex smoothly catalyzes a base-free nitroaldol condensation in a highly enantioselective manner (ee up to 93%) also in reagent-grade solvent and in the presence of air. Effective recovery and recycling (up to five runs) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service