Skip to Content
Merck
All Photos(3)

Key Documents

D110000

Sigma-Aldrich

3,5-Dihydroxybenzoic acid

97%

Synonym(s):

α-Resorcylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
2207864
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

236-238 °C (dec.) (lit.)

SMILES string

OC(=O)c1cc(O)cc(O)c1

InChI

1S/C7H6O4/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3,8-9H,(H,10,11)

InChI key

UYEMGAFJOZZIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seetharaman Vaidyanathan et al.
Rapid communications in mass spectrometry : RCM, 20(8), 1192-1198 (2006-03-17)
Matrix-assisted laser desorption/ionisation (MALDI) mass spectrometry was investigated for the simultaneous detection of several metabolites, as applicable to global metabolite analysis (metabolomics). The commonly employed organic matrices alpha-cyano-4-hydroxycinnamic acid and 3,5-dihydroxybenzoic acid, in both the crystalline and ionic liquid forms
Abigail E Wolfe et al.
Biochemistry, 46(19), 5741-5753 (2007-04-21)
Dihydroorotate dehydrogenases (DHODs) catalyze the oxidation of dihydroorotate to orotate in the only redox reaction in pyrimidine biosynthesis. The pyrimidine binding sites are very similar in all structurally characterized DHODs, suggesting that the prospects for identifying a class-specific inhibitor directed
Silke C Wenzel et al.
Chembiochem : a European journal of chemical biology, 9(16), 2711-2721 (2008-10-31)
Kendomycin is a bioactive polyketide that is produced by various Streptomyces strains. It displays strong antibiotic activities against a wide range of bacteria and exhibits remarkable cytotoxic effects on the growth of several human cancer cell lines. In this study
Anja Koskela et al.
Journal of agricultural and food chemistry, 56(17), 7678-7681 (2008-08-12)
This study presents the optimization and validation of a rapid protocol for quantifying alkyresorcinol (AR) metabolites 3,5-dihydroxybenzoic acid (DHBA) and 3-(3,5-dihydroxyphenyl)-1-propanoic acid (DHPPA) in plasma, using high-performance liquid chromatography (HPLC) coupled with a coulometric electrode array detector. Syringic acid (SyrA)
Qian Cao et al.
Biosensors & bioelectronics, 25(12), 2680-2685 (2010-06-01)
A colorimetric, label-free, and nonaggregation-based gold nanoparticles probe has been developed for the detection of melamine. Gold nanoparticles were generated using 3,5-dihydroxybenzoic acid as a reducer without adding gold nanoparticle seeds at room temperature. Upon the addition of melamine, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service