I2304
1-Indanone
ReagentPlus®, ≥99%
Synonym(s):
1-Oxoindane, α-Hydrindone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H8O
CAS Number:
Molecular Weight:
132.16
Beilstein:
507957
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
product line
ReagentPlus®
Assay
≥99%
bp
243-245 °C (lit.)
mp
38-40 °C (lit.)
density
1.103 g/mL at 25 °C (lit.)
SMILES string
O=C1CCc2ccccc12
InChI
1S/C9H8O/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4H,5-6H2
InChI key
QNXSIUBBGPHDDE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1-Indanone can be used to synthesize:
It can also be used to build other indan-based systems like truxene, prekinamycin, indan-C60 and biindanylidenes.
- 11H-Indeno-[1,2-b]-quinolin-10-ylamine, a potential acetylcholinesterase inhibitor for the treatment of Alzheimer′s disease.
- CoumBARAC (coumarin fused biarylazacyclooctynone) for fluorogenic real-time imaging of azide-labeled biomolecules.
- Light emitting, organic semiconductors such as 4-pyrenyl-2-piperidin-1-yl-9H-fluorene-1-carbonitrile and bisindenoanthrazolines for organic light emitting diode (OLED) applications.
It can also be used to build other indan-based systems like truxene, prekinamycin, indan-C60 and biindanylidenes.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
233.0 °F - closed cup
Flash Point(C)
111.67 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Trisannulated benzene derivatives by acid catalyzed aldol cyclotrimerizations of cyclic ketones. Methodology development and mechanistic insight.
Amick AW and Scott LT.
The Journal of Organic Chemistry, 72(9), 3412-3418 (2007)
Biindanylidenes: role of central bond torsion in nonvertical triplet excitation transfer to the stilbenes.
Saltiel J, et al.
Journal of the American Chemical Society, 125(52), 16158-16159 (2003)
Novel n-type conjugated ladder heteroarenes: Synthesis, self-assembly of nanowires, electron transport, and electroluminescence of bisindenoanthrazolines.
Ahmed E, et al.
Chemistry of Materials, 22(20), 5786-5796 (2010)
Indan-C 60: from a crystalline molecule to photovoltaic application.
Wang T, et al.
Chemical Communications (Cambridge, England), 49(85), 9923-9925 (2013)
Highly efficient phosphorescent light-emitting diodes by using an electron-transport material with high electron affinity.
Earmme T, et al.
The Journal of Physical Chemistry C, 113(43), 18448-18450 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service