Skip to Content
Merck
All Photos(2)

Key Documents

117455

Sigma-Aldrich

p-Toluenesulfonyl fluoride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2F
CAS Number:
Molecular Weight:
174.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

reaction suitability

reaction type: click chemistry

bp

112 °C/16 mmHg (lit.)

mp

41-42 °C (lit.)

SMILES string

Cc1ccc(cc1)S(F)(=O)=O

InChI

1S/C7H7FO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3

InChI key

IZZYABADQVQHLC-UHFFFAOYSA-N

General description

p-Toluenesulfonyl fluoride is a peroxygen bleach activator and is also useful in the treatment of Alzheimer′s disease.

Application

p-Toluenesulfonyl fluoride was used to study the DNS polymerase from spinach leaf extracts. It was used in isolation and separation of Pseudomonas aeruginosa membranes by density sucrose gradient centrifugation.
Protease inhibitor.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinkui Yang et al.
Biotechnology letters, 27(15), 1123-1128 (2005-09-01)
Lecanicillium psalliotae produced an extracellular protease (Ver112) which was purified to apparent homogeneity giving a single band on SDS-PAGE with a molecular mass of 32 kDa. The optimum activity of Ver112 was at pH 10 and 70 degrees C (over
Nobuyuki Shibata et al.
FEMS microbiology letters, 256(1), 137-144 (2006-02-21)
Lipophilic Malassezia species may induce catheter-associated sepsis in premature neonates and immunocompromised patients receiving parenteral lipid emulsions. To assess the participation of lipolytic enzymes in the pathogenesis of this yeast, we cloned a gene encoding the enzyme. A lipolytic enzyme
Tsuyoshi Fujita et al.
Cellular signalling, 17(2), 167-175 (2004-10-21)
In this study, we examined the mechanism by which CD38 cleavage is regulated through the mitogen-activated protein (MAP) kinases after stimulation by fMLP and interleukin-8 (IL-8) in neutrophils. Both fMLP and IL-8 increased chemotaxis and decreased CD38 protein in neutrophils
Tomonori Kaneko et al.
Protein science : a publication of the Protein Society, 14(2), 558-565 (2005-01-06)
Growth-limiting stresses in bacteria induce the general stress response to protect the cells against future stresses. Energy stress caused by starvation conditions in Bacillus subtilis is transmitted to the sigma(B) transcription factor by stress-response regulators. RsbP, a positive regulator, is
Novelties of solid-liquid phase transfer catalysed synthesis of p-toluenesulfonyl fluoride from p-toluenesulfonyl chloride.
Yadav GD and Paranjape PM.
Journal of Fluorine Chemistry, 126(3), 289-295 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service