Skip to Content
Merck
All Photos(1)

Key Documents

282332

Sigma-Aldrich

Benzenesulfonyl fluoride

99%

Synonym(s):

Phenylsulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2F
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%
Pricing and availability is not currently available.

Quality Level

Assay

99%

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.492 (lit.)

bp

207-208 °C (lit.)

density

1.333 g/mL at 25 °C (lit.)

SMILES string

FS(=O)(=O)c1ccccc1

InChI

1S/C6H5FO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H

InChI key

IDIPWEYIBKUDNY-UHFFFAOYSA-N

General description

The gas-phase fluorine migration reactions of radical cations of benzenesulfonyl fluoride have been studied by electron ionization tandem mass spectrometry[1].

Application

Benzenesulfonyl fluoride has been used in the preparation of α-sulfonyl phosphonates by direct sulfonylation of lithiated alkyl phosphonates[2].

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Ren et al.
Aging cell, 16(5), 976-987 (2017-07-07)
Aging is accompanied with unfavorable geometric and functional changes in the heart involving dysregulation of Akt and autophagy. This study examined the impact of Akt2 ablation on life span and cardiac aging as well as the mechanisms involved with a
Jael Quintero et al.
Parasitology international, 66(3), 324-330 (2017-01-23)
Humoral and cellular immune responses play an important role during Giardia lamblia infection. Several Giardia proteins have been identified as immunogenic antigens based on their elicited humoral immune response. Poorly is known about Giardia antigens that stimulate a cellular immune
Lilah Glazer et al.
Journal of proteomics, 128, 333-343 (2015-09-01)
Chitin is a major component of arthropod cuticles, where it forms a three-dimensional network that constitutes the scaffold upon which cuticles form. The chitin fibers that form this network are closely associated with specific structural proteins, while the cuticular matrix
Direct Sulfonylation of Lithiated Alkyl Phosphonates with Benzenesulfonyl Fluoride; Facile Method for Preparation of a-Sulfonyl Alkyl Phosphonates and Vinyl Sulfones.
Jang WB, et al.
Synthetic Communications, 28(7), 1253-1256 (1998)
Steffen Mayerl et al.
Endocrinology, 156(2), 755-767 (2014-12-10)
TRH not only functions as a thyrotropin releasing hormone but also acts as a neuropeptide in central circuits regulating food intake and energy expenditure. As one suggested mode of action, TRH expressed in the caudal brainstem influences vagal activity by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service