Skip to Content
Merck
All Photos(2)

Key Documents

115495

Sigma-Aldrich

4,4′-Difluorobenzophenone

99%

Synonym(s):

Bis(4-fluorophenyl) ketone, Bis(4-fluorophenyl)methanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(FC6H4)2CO
CAS Number:
Molecular Weight:
218.20
Beilstein:
516231
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

102-105 °C (lit.)

functional group

fluoro
ketone

SMILES string

Fc1ccc(cc1)C(=O)c2ccc(F)cc2

InChI

1S/C13H8F2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8H

InChI key

LSQARZALBDFYQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4′-Difluorobenzophenone has been used in the study of thermal stability, water uptake and proton conductivity of carboxylated, carboxylated/sulfonated, and crosslinked membranes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - Pensky-Martens closed cup

Flash Point(C)

152 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Poly (aryl ether ketone) s with carboxylic acid groups: synthesis, sulfonation and crosslinking.
Liu B, et al.
Journal of Materials Chemistry, 18(39), 4675- 4682 (2008)
Sinan Feng et al.
ACS applied materials & interfaces, 9(29), 24527-24537 (2017-07-07)
A novel semicrystalline poly(ether ketone) (PEK)-based proton exchange membrane (semi-SPEK-x) has been developed. Through a one-step sulfonation and hydrolysis, a poly(ether ketimine) precursor transforms into PEK and ion-conducting groups are introduced. With an ion-exchange capacity ranging from 1.49 to 2.00
S Kariya et al.
Research communications in chemical pathology and pharmacology, 78(1), 85-95 (1992-10-01)
The oxidative metabolism of flunarizine [1-[bis(4-fluorophenyl)-methyl]-4-(3-phenyl-2-propenyl)piperazine, FZ] to 1-[bis-(4-fluorophenyl)methyl]piperazine (M-1), 1-[bis(4-fluorophenyl)methyl]-4-[3-(4'-hydroxyphenyl)-2- propenyl]piperazine (M-2) and 4,4'-difluorobenzophenone (M-3) has been studied in liver microsomes of Wistar and Dark Agouti (DA) rats. Kinetic analysis demonstrated a sex difference (male > female) in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service