Skip to Content
Merck
All Photos(1)

Key Documents

W312908

Sigma-Aldrich

Biphenyl

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H5
CAS Number:
Molecular Weight:
154.21
FEMA Number:
3129
Beilstein:
1634058
EC Number:
Council of Europe no.:
10978
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.013
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

Agency

meets purity specifications of JECFA

vapor density

5.31 (vs air)

vapor pressure

9.46 mmHg ( 115 °C)

Assay

≥99%

form

flakes
powder or crystals

autoignition temp.

1004 °F

expl. lim.

0.6 %, 111 °F
5.8 %, 166 °F

bp

255 °C (lit.)

mp

68-70 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

geranium; green

SMILES string

c1ccc(cc1)-c2ccccc2

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Biphenyl is an aromatic hydroacarbon that is generally used as a preservative for citrus fruits.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Drazenka Selesi et al.
FEMS microbiology ecology, 68(1), 86-93 (2009-02-04)
The aromatic hydrocarbon biphenyl is a widely distributed environmental pollutant. Whereas the aerobic degradation of biphenyl has been extensively studied, knowledge of the anaerobic biphenyl-oxidizing bacteria and their biochemical degradation pathway is scarce. Here, we report on an enrichment culture
Sheng-Hao Hsu et al.
Acta biomaterialia, 8(11), 4151-4161 (2012-07-31)
Low-shrinkage resin-based photocurable liquid crystalline epoxy nanocomposite has been investigated with regard to its application as a dental restoration material. The nanocomposite consists of an organic matrix and an inorganic reinforcing filler. The organic matrix is made of liquid crystalline
Shi-Rong Liu et al.
Organic letters, 15(4), 878-881 (2013-02-05)
A BODIPY-based fluorescent probe, HCSe, has been successfully developed for the rapid detection of hypochlorous acid based on the specific HOCl-promoted oxidation of diphenyl selenide in response to the amount of HOCl. Confocal fluorescence microscopy imaging using RAW264.7 cells showed
Xiaofeng Chen et al.
The Journal of organic chemistry, 77(17), 7572-7578 (2012-08-07)
A highly efficient cross-coupling of diarylborinic acids and anhydrides with aryl chlorides and bromides has been effected by using a palladium catalyst system co-supported by a strong σ-donor N-heterocyclic carbene (NHC), N,N'-bis(2,6-diisopropylphenyl) imidazol-2-ylidene, and a strong π-acceptor phosphite, triphenylphosphite, in
Susan L Zultanski et al.
Journal of the American Chemical Society, 135(2), 624-627 (2013-01-04)
The first Suzuki cross-couplings of unactivated tertiary alkyl electrophiles are described. The method employs a readily accessible catalyst (NiBr(2)·diglyme/4,4'-di-tert-butyl-2,2'-bipyridine, both commercially available) and represents the initial example of the use of a group 10 catalyst to cross-couple unactivated tertiary electrophiles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service