Skip to Content
Merck
All Photos(1)

Documents

10851

Sigma-Aldrich

Kuraray Poval ®56-98

Mw ~195,000

Synonym(s):

Poly(vinyl alcohol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

mol wt

Mw ~195,000

extent of labeling

98.0-98.8 mol% hydrolysis
~4,300 polymerization

ign. residue

≤1%

viscosity

52-60 mPa.s, 4 % in H2O(20 °C)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Mowiol is a registered trademark of Kuraray Europe GmbH

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT SE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yani Guo et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 48(5), 518-525 (2013-02-07)
The degradation characteristics of the new mixed strains isolated from a de-sizing process which could degrade PVA thoroughly were studied. The interrelation of seven kinds of single bacterium separated from mixed strains was studied through combination experiments. The degradation process
Fusako Kawai et al.
Archives of microbiology, 195(2), 131-140 (2012-12-25)
Scanning electron microscopy (SEM) shows remarkable morphological surface changes in Sphingopyxis sp. 113P3 cells grown in polyvinyl alcohol (PVA) but not in Luria-Bertani medium (LB) (Hu et al. in Arch Microbiol 188: 235-241, 2007). However, transmission electron microscopy showed no
Jie Xue et al.
Journal of photochemistry and photobiology. B, Biology, 119, 31-36 (2013-01-15)
Synthetic adhesives inspired by marine mussel have attracted great attention due to its excellent water-resistance and good biocompatibility. In this study, a photocrosslinkable bioadhesive containing 3,4-Dihydroxy-l-phenylalanine (DOPA) functional group, which is central to curing mussel adhesive proteins, was prepared by
Cijun Shuai et al.
Biofabrication, 5(1), 015014-015014 (2013-02-07)
A tetragonal polyvinyl alcohol (PVA) scaffold with 3D orthogonal periodic porous architecture was fabricated via selective laser sintering (SLS) technology. The scaffold was fabricated under the laser power of 8 W, scan speed of 600 mm min(-1), laser spot diameter of 0.8 mm
Feng-yong Liu et al.
Acta radiologica (Stockholm, Sweden : 1987), 54(1), 61-66 (2013-02-05)
Preoperative embolization of tumors is a well-established procedure that has been successfully applied in various clinical situations. Preoperative embolization can reduce the vascularity of tumors resulting in a clearer operative field, less difficult dissection, decreased blood loss, and, in some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service