Skip to Content
Merck
All Photos(2)

Documents

563900

Sigma-Aldrich

Poly(vinyl alcohol)

average Mw 130,000, 99+% hydrolyzed

Synonym(s):

PVA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

form

powder or crystals

mol wt

average Mw 130,000

falling ball

56-68 cP, 4 % in H2O(20 °C)(lit.)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl alcohol) (PVA) is a vinyl based water soluble polymer that can be used as a non-ionic surface active agent. It can also be used as a biodegradable polymer and can be incorporated in adhesives, coatings, textiles, ceramics and cosmetics. Its properties include good transparency, chemical resistance, toughness and anti-electrostatic properties.

Application

PVA can be used in a variety of applications such as:
  • a blended membrane for separation of gases
  • a proton conducting membrane for the development of fuel cells
  • hydrogels in tissue engineering

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compostable Polymer Properties and Packaging Applications
Plastic Films in Food Packaging, 217-248 (2013)
Relationship between micro-porosity, water permeability and mechanical behavior in scaffolds for cartilage engineering
Vikingsson L, et al.
Journal of the Mechanical Behavior of Biomedical Materials, 48(2), 60-69 (2015)
Plastics Materials, 386-397 (1999)
Cross-linked poly (vinyl alcohol)/sulfonated nanoporous silica hybrid membranes for proton exchange membrane fuel cell
Beydaghi H, et al.
Journal of Nanostructure in Chemistry, 4(2), 97-97 (2014)
CO2 separation using thermally stable crosslinked poly (vinyl alcohol) membrane blended with polyvinylpyrrolidone/polyethyleneimine/tetraethylenepentamine
Mondal A and Mandal B
Journal of Membrane Science , 460(6), 126-138 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service