Merck
  • Biodegradation of PVA by the new mixed strains isolated from a de-sizing process.
Biodegradation of PVA by the new mixed strains isolated from a de-sizing process.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2013-02-07)
Yani Guo, Ming Zhou, Shuang K Cui, Ning Nian
ABSTRACT
The degradation characteristics of the new mixed strains isolated from a de-sizing process which could degrade PVA thoroughly were studied. The interrelation of seven kinds of single bacterium separated from mixed strains was studied through combination experiments. The degradation process was analysed through UV spectrum, IR, GPC and other methods. The enzymatic properties and enzymatic reaction dynamic processes were studied. The results showed that PVA with the initial average molecular weight of 51,260 Da could be degraded thoroughly by seven kinds of single bacterium because of their mutual coordination effect. In addition, some carboxylic acid and methyl ketone substances could be produced in the PVA degradation process by some bacteria (mainly A4 and C1). These middle smaller molecular weight materials would be further degraded thoroughly by other bacteria (mainly A1 and A3). The PVA-degrading enzyme that was produced by the mixed strains was mainly an exocellular enzyme. The enzyme reaction kinetics equation was v = 19.5[S]/(2.06 × 10(-3) + [S]).
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000