Skip to Content
Merck
All Photos(1)

Key Documents

W390909

Sigma-Aldrich

Cyclohexanone

99.8%

Synonym(s):

Hexanon, Keto hexamethylene, Ketohexamethylene, Pimelic ketone, Sextone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14
FEMA Number:
3909
Beilstein:
385735
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.148
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Agency

meets purity specifications of JECFA

vapor density

3.4 (vs air)

vapor pressure

3.4 mmHg ( 20 °C)

Assay

99.8%

form

liquid

autoignition temp.

788 °F

expl. lim.

1.1 %, 100 °F
9.4 %

refractive index

n20/D 1.450 (lit.)

bp

155 °C (lit.)

mp

−47 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

minty

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cyclohexanone is a cyclic ketone with a minty odor. It is reported to be present in volatile flavor fraction of kiwi fruit pulp and acerola fruit.

Application


  • Structural Revision of Hyperibrin B and Hyperscabrones H and I by Biosynthetic Considerations, NMR Analysis, and Chemical Synthesis.: Cyclohexanone is utilized in the synthesis and structural revision of complex natural products, demonstrating its utility in biochemical research (Wang et al., 2021).

  • Biocatalytic conversion of cycloalkanes to lactones using an in-vivo cascade in Pseudomonas taiwanensis VLB120.: This study highlights the biocatalytic potential of Cyclohexanone in producing valuable lactones, contributing to advancements in green chemistry (Karande et al., 2018).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GC-FTIR as a powerful tool for the characterization of flavor components in Kiwi.
Fischbock G, et al.
Microchimica Acta, 96(1), 249-257 (1988)
Application of fragment screening and merging to the discovery of inhibitors of the Mycobacterium tuberculosis cytochrome P450 CYP121.
Sean A Hudson et al.
Angewandte Chemie (International ed. in English), 51(37), 9311-9316 (2012-08-15)
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2629-2637 (2012-09-07)
Iron (and to a lesser extent manganese) in the wall of a 316 stainless steel (SS) reactor is responsible for the hydrogenation of cyclohexanone to cyclohexanol when using an aqueous formic acid solution under high temperature and pressure water (HTPW)
Alakananda Hajra et al.
Organic letters, 14(21), 5488-5491 (2012-10-18)
Dehydrogenative aromatization of cyclohexanone imines to arylamines has been achieved using a palladium catalyst under aerobic conditions. The reaction is applicable to a variety of imines that are either preformed or generated in situ from cyclohexanone derivatives and aryl or
Brahm J Yachnin et al.
Journal of the American Chemical Society, 134(18), 7788-7795 (2012-04-18)
The Baeyer-Villiger monooxygenases (BVMOs) are a family of bacterial flavoproteins that catalyze the synthetically useful Baeyer-Villiger oxidation reaction. This involves the conversion of ketones into esters or cyclic ketones into lactones by introducing an oxygen atom adjacent to the carbonyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service