Skip to Content
Merck
All Photos(1)

Key Documents

105481

Sigma-Aldrich

1,3-Diphenylisobenzofuran

97%

Synonym(s):

1,3-Diphenyl-2-benzofuran, 2,5-Diphenyl-3,4-benzofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14O
CAS Number:
Molecular Weight:
270.32
Beilstein:
199922
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

128-130 °C (lit.)

functional group

phenyl

SMILES string

c1ccc(cc1)-c2oc(-c3ccccc3)c4ccccc24

InChI

1S/C20H14O/c1-3-9-15(10-4-1)19-17-13-7-8-14-18(17)20(21-19)16-11-5-2-6-12-16/h1-14H

InChI key

ZKSVYBRJSMBDMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Diphenylisobenzofuran is a fluorescent dye[1].1,3-Diphenylisobenzofuran is the model compound in studies of singlet fission[2].

Application

1,3-Diphenylisobenzofuran(DPBF) was used as a fluorescent probe for detection of superoxide anion radical (O2) inside the membrane lipid layer by DPBF fluorescence quenching method[1]. 1,3-Diphenylisobenzofuran(DPBF) was used as quencher during the photoinactivation of TA-3 mouse mammary carcinoma cells containing hematoporphyrin[3]. 1,3-Diphenylisobenzofuran(DPBF) was used to study the single crystal molecular structure and solution photophysical properties of DPBF[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Weishaupt et al.
Cancer research, 36(7 PT 1), 2326-2329 (1976-07-01)
Singlet oxygen, a metastable state of normal triplet oxygen, has been identified as the cytotoxic agent that is probably responsible for in vitro inactivation of TA-3 mouse mammary carcinoma cells following incorporation of hematoporphyrin and exposure to red light. This
Meng Duan et al.
Nanoscale, 11(39), 18426-18435 (2019-10-03)
This work explored the application of matrix metalloproteinase 2-targeted superparamagnetic nanoprobes for magnetic resonance imaging (MRI), near infrared (NIR) fluorescence imaging and photodynamic therapy of tumors. PEG, PAMAM (G5) and matrix metalloproteinase 2 (MMP2) were attached to the surface of
Diana Zaharie-Butucel et al.
Journal of colloid and interface science, 552, 218-229 (2019-05-28)
Significant efforts are currently being funneled into the improvement of therapeutic outcomes in cancer by designing hybrid nanomaterials that synergistically combine chemotherapeutic abilities and near-infrared (NIR) light-activated photothermal (PTT) and photodynamic (PDT) activity. Herein, a nanotherapeutic platform is specifically designed
T Ohyashiki et al.
Biochimica et biophysica acta, 1421(1), 131-139 (1999-11-16)
Utilization of a fluorescence dye, 1,3-diphenylisobenzofuran (DPBF) as a detector of superoxide anion radical (O2*-) was examined. The fluorescence intensity of DPBF incorporated in phospholipid liposomes consisting of phosphatidylcholine (PC) and phosphatidylserine (PS) is effectively quenched by incubation with xanthine/xanthine
Jia Wen et al.
Scientific reports, 6, 38931-38931 (2016-12-13)
In this work, a triple-stimuli (GSH, pH and light irradiation) responsive system were designed based on CeO2 nanoparticles (CeO2 NPs) coated doxorubicin (DOX) and photosensitizer hematoporphyrin (HP) dual-loaded mesoporous silica nanoparticles (MSN). Upon entering into cancer cells, both high concentration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service