Skip to Content
Merck
All Photos(1)

Key Documents

102644

Sigma-Aldrich

Ethyl 4-fluorobenzoate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CO2C2H5
CAS Number:
Molecular Weight:
168.16
Beilstein:
1866794
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.486 (lit.)

bp

210 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

CCOC(=O)c1ccc(F)cc1

InChI

1S/C9H9FO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2H2,1H3

InChI key

UMPRJGKLMUDRHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 4-fluorobenzoate is a clear colorless liquid. The purified [18F]fluoride is used for solution phase nucleophilic labeling of ethyl [18F]4-fluorobenzoate. Ethyl 4-fluorobenzoate causes N-arylation of 5-methoxyindole in presence of 18-crown-6 by using microwave-assisted technology.

Application

Ethyl 4-fluorobenzoate may be used in the synthesis of 4,4′-(1,4-piperazinediyl) bisbenzoic acid ethyl ester.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Martínez-Bueno et al.
Plasmid, 28(1), 61-69 (1992-07-01)
Production of bacteriocin Bc-48 by Enterococcus faecalis S-48 is encoded by the conjugative plasmid pMB1, which is approximately 90 kb and also responds to sex pheromones of E. faecalis OG1X. Mutants harboring deleted forms of this plasmid (pMB1-del, 75 kb)
Investigation of the N-arylation of various substituted indoles using microwave-assisted technology.
Frayne GL and Green GM.
Tetrahedron Letters, 49(51), 7328-7329 (2008)
G K Mulholland
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes, 42(11), 1003-1008 (1991-01-01)
No-carrier-added trimethylsilyl[18F]fluoride (TMS[18F]F) was rapidly liberated from a variety of dry supports containing unreactive [18F]fluoride by simply heating with neat bistrimethylsilylsulfate. The supports included calcium phosphate, anion exchange resins, alumina, borosilicate and porous carbon beads, to which [18F]fluoride was applied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service