Skip to Content
Merck
All Photos(4)

Key Documents

330000

Sigma-Aldrich

Rose bengal

Dye content 95 %, ≥96% (HPLC)

Synonym(s):

4,5,6,7-Tetrachloro-2′,4′,5′,7′-tetraiodofluorescein disodium salt, Acid Red 94, Bengal Rose B sodium salt, Rose Bengal sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H2Cl4I4Na2O5
CAS Number:
Molecular Weight:
1017.64
Beilstein:
3645857
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥96% (HPLC)

form

solid

composition

Dye content, 95%

technique(s)

titration: suitable

color

red to brown

ε (extinction coefficient)

≥ 3000 at 350-354 nm
≥12000 at 311-315 nm
≥32000 at 262-266 nm
≥32000 at 513-517 nm
≥98000 at 546-550 nm at 0.04 g/L (H2O + 1ml 1% Na2CO3)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]C(=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1C2=C3C=C(I)C(=O)C(I)=C3Oc4c(I)c([O-])c(I)cc24

InChI

1S/C20H4Cl4I4O5.2Na/c21-10-8(9(20(31)32)11(22)13(24)12(10)23)7-3-1-5(25)16(29)14(27)18(3)33-19-4(7)2-6(26)17(30)15(19)28;;/h1-2,29H,(H,31,32);;/q;2*+1/p-2

InChI key

UWBXIFCTIZXXLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Rose Bengal is a red or rose color dye and is anionic in nature. It is a 4,5,6,7-tetrachloro 2′,4′,5′,7′-tetraiodo derivative of fluorescein. It is widely used to detect damage to the ocular surface epithelium in ocular surface diseases such as dry eye and herpetic keratitis. Rose Bengal has mild antiviral potency.
Rose Bengal is used to stain dead and degenerating cells and mucus.

Application

Rose Bengal has been used to evaluate acrosome integrity. It has also been used to initiate photothrombotic occlusion in blood.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

New insights for male infertility revealed by alterations in spermatic function and differential testicular expression of thyroid-related genes
Romano RM, et al.
Endocrine, 55(2), 607-617 (2017)
Photothrombotic induction of capillary ischemia in the mouse cortex during in vivo two-photon imaging
Underly RG and Shih AY
Bio-protocol, 7(13) (2017)
Brian Duvall, Robert M. Kershner
Ophthalmic Medications and Pharmacology (2006)
The hypothalamic-pituitary-testicular axis and the testicular function are modulated after silver nanoparticle exposure
Cavallin MD, et al.
Toxicology Research, 7(1), 102-116 (2018)
Mucin Characteristics of Human Corneal-Limbal Epithelial Cells that Exclude the Rose Bengal Anionic Dye
Argueso P
Investigative Ophthalmology & Visual Science, 47, 113-119 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service