Merck
  • FOXM1-LDHA signaling promoted gastric cancer glycolytic phenotype and progression.
FOXM1-LDHA signaling promoted gastric cancer glycolytic phenotype and progression.
International journal of clinical and experimental pathology (2015-08-12)
Weihua Jiang, Fei Zhou, Ning Li, Qi Li, Liwei Wang
ABSTRACT
The oncogenic transcription factor forkhead box protein M1 (FOXM1) plays critical roles in gastric cancer (GC) development and progression. However, the underlying mechanisms has not fully demonstrated. Lactate dehydrogenase A (LDHA) is widely overexpressed in a series of cancers and is one of the two subunits of Lactate dehydrogenase (LDH), which is the key glycolytic enzyme and catalyzes the interconversion of pyruvate and lactate. In this study, we characterized the regulation of aerobic glycolysis by FOXM1 via transactivation of LDHA in GC. We found that LDHA was overexpressed GC cells, and the expression of LDHA was transcriptionally regulated by FOXM1. Furthermore, FOXM1 regulated GC cells glycolytic phenotype, proliferation, migration and invasion via LDHA. Thus, FOXM1-LDHA signaling functioned as a stimulator of glycolysis and promoted GC progression.
Sigma-Aldrich
MISSION® esiRNA, targeting human FOXM1
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Foxm1
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Sodium oxamate, ≥98%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)