Skip to Content
Merck
All Photos(2)

Documents

G5146

Sigma-Aldrich

D-(+)-Glucose

Hybri-Max, powder, BioReagent, suitable for hybridoma

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.75

biological source

maize

grade

Hybri-Max

product line

BioReagent

Assay

≥99.5%

form

powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

mp

150-152 °C (lit.)

solubility

H2O: 2g + 15 mL (Tested)

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

General description

Glycogen phosphorylase (PYG) exists as brain and muscle isoform. It is expressed in astrocytes and neurons respectively. The PYGM gene is mapped to human chromosome 11q13.1.

Application

D-(+)-Glucose has been used in cell engineering. It has also been used to perform oral glucose tolerance test.

Biochem/physiol Actions

Glycogen phosphorylase, muscle associated (PYGM), is an important contributor to glycogenolysis. Down regulation of PYGM gene is observed in schizophrenia. Mutation in PYGM leads to McArdle disease, a glycogen storage disorder. The PYGM gene is significantly associated with energy production.

Preparation Note

The material may produce a yellowish solution if prepared at a higher concentration than 2g + 15ml.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic evaluation of AMPD1, CPT2, and PGYM metabolic enzymes in patients with chronic fatigue syndrome
Maltese PE, et al.
Genetics and molecular research : GMR, 15(3), 1-10 (2016)
Novel variant in the PYGM gene causing late-onset limb-girdle myopathy, ptosis, and camptocormia
Cheraud C, et al.
Muscle and Nerve, 57(1), 157-160 (2018)
Electrical vs manual acupuncture stimulation in a rat model of polycystic ovary syndrome: different effects on muscle and fat tissue insulin signaling
Johansson J, et al.
PLoS ONE, 8(1) (2013)
Comparative metabolic analysis of CHO cell clones obtained through cell engineering, for IgG productivity, growth and cell longevity
Wilkens CA and Gerdtzen ZP
PLoS ONE, 10(3) (2015)
CDKN1C/p57 kip2 is a candidate tumor suppressor gene in human breast cancer
Larson PS, et al.
BMC Cancer, 8(1), 68-68 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service