Merck
  • STAT5 programs a distinct subset of GM-CSF-producing T helper cells that is essential for autoimmune neuroinflammation.
STAT5 programs a distinct subset of GM-CSF-producing T helper cells that is essential for autoimmune neuroinflammation.
Cell research (2014-11-22)
Wanqiang Sheng, Fan Yang, Yi Zhou, Henry Yang, Pey Yng Low, David Michael Kemeny, Patrick Tan, Akira Moh, Mark H Kaplan, Yongliang Zhang, Xin-Yuan Fu
ABSTRACT
T helper (TH)-cell subsets, such as TH1 and TH17, mediate inflammation in both peripheral tissues and central nervous system. Here we show that STAT5 is required for T helper-cell pathogenicity in autoimmune neuroinflammation but not in experimental colitis. Although STAT5 promotes regulatory T cell generation and immune suppression, loss of STAT5 in CD4+ T cells resulted in diminished development of experimental autoimmune encephalomyelitis (EAE), a mouse model of multiple sclerosis. Our results showed that loss of encephalitogenic activity of STAT5-deficient autoreactive CD4+ T cells was independent of IFN-γ or interleukin 17 (IL-17) production, but was due to the impaired expression of granulocyte-macrophage colony-stimulating factor (GM-CSF), a crucial mediator of T-cell pathogenicity. We further showed that IL-7-activated STAT5 promotes the generation of GM-CSF-producing CD4+ T cells, which were preferentially able to induce more severe EAE than TH17 or TH1 cells. Consistent with GM-CSF-producing cells being a distinct subset of TH cells, the differentiation program of these cells was distinct from that of TH17 or TH1 cells. We further found that IL-3 was secreted in a similar pattern as GM-CSF in this subset of TH cells. In conclusion, the IL-7-STAT5 axis promotes the generation of GM-CSF/IL-3-producing TH cells. These cells display a distinct transcriptional profile and may represent a novel subset of T helper cells which we designate as TH-GM.
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, purum, ≥99.0% (GC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)