Merck
  • Chemical and sensory effects of storing sauvignon Blanc wine in colored bottles under artificial light.
Chemical and sensory effects of storing sauvignon Blanc wine in colored bottles under artificial light.
Journal of agricultural and food chemistry (2014-07-02)
Alejandro Cáceres-Mella, Daniela Flores-Valdivia, V Felipe Laurie, Remigio López-Solís, Álvaro Peña-Neira
ABSTRACT
The chemical and sensory effects of storing Sauvignon Blanc in colored bottles and exposing them to artificial light were examined. The colors of the bottles chosen were Dead Leaf Green, Antique Green, Amber, and Flint. The light was provided by fluorescent tubes with a regime of 16 h of exposure during 8 months of storage. The results indicated that the wine's chemical composition was affected by the type of bottle used. The Flint bottle presented the lowest concentration of total phenols. Yellow coloration was not dependent on the bottle color, as the wine in darker bottles (Amber, Antique Green, and Dead Leaf Green) had considerably more yellow color development than the wine in clear bottles. With regard to the sensory analyses performed, a trend showing an increase in color intensity and a decrease in overall aromas depending on the bottle color was observed. The wine's aromatic description changed significantly during its storage under artificial light conditions, demonstrating a decrease in vegetal aromas and an increase in citrus and tropical flavors that was dependent on the bottle color.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Caftaric acid, ≥97.0%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Caftaric acid, analytical standard
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Supelco
Acetic acid, analytical standard
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)