Skip to Content
Merck
All Photos(8)

Documents

793531

Sigma-Aldrich

Sodium sulfate

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

product line

ReagentPlus®
Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

loss

≤0.5% loss on drying

pH

5.2-8 (20 °C, 50 g/L)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Iron Phosphate Purification: Sodium sulfate is involved in methods for removing impurities from chemical compounds. A recent study focused on developing a technique to remove sodium sulfate from iron phosphate, which is crucial for producing high-purity materials for various industrial applications. This research highlights sodium sulfate′s role in refining chemical processes and improving product quality (Li et al., Advances in Analytical Chemistry, 2022).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teppei Adachi et al.
Inflammatory bowel diseases, 21(1), 31-39 (2014-12-02)
Expression of heat shock protein A4 (HSPA4, also called Apg-2), a member of the HSP110 family, is induced by several forms of stress. The physiological and pathological functions of HSPA4 in the intestine remain to be elucidated. We assessed HSPA4
Christoph Zürcher et al.
Journal of virology, 88(13), 7235-7243 (2014-04-18)
The RNase activity of the envelope glycoprotein E(rns) of the pestivirus bovine viral diarrhea virus (BVDV) is required to block type I interferon (IFN) synthesis induced by single-stranded RNA (ssRNA) and double-stranded RNA (dsRNA) in bovine cells. Due to the
Rebecca L Scalzo et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(6), 2705-2714 (2014-03-07)
Improved endurance exercise performance in adult humans after sprint interval training (SIT) has been attributed to mitochondrial biogenesis. However, muscle protein synthesis (MPS) and mitochondrial biogenesis during SIT have not been measured, nor have sex-specific differences. We hypothesized that males
Roosmarijn E Vandenbroucke et al.
Nature reviews. Drug discovery, 13(12), 904-927 (2014-11-08)
Matrix metalloproteinases (MMPs) are zinc-dependent endopeptidases that form a family of 24 members in mammals. Evidence of the pathological roles of MMPs in various diseases, combined with their druggability, has made them attractive therapeutic targets. Initial drug discovery efforts focused
Erik C Dreaden et al.
ACS nano, 8(8), 8374-8382 (2014-08-08)
Active targeting of nanoscale drug carriers can improve tumor-specific delivery; however, cellular heterogeneity both within and among tumor sites is a fundamental barrier to their success. Here, we describe a tumor microenvironment-responsive layer-by-layer (LbL) polymer drug carrier that actively targets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service