Merck
  • Durable diagnosis of seminal vesicle and sexual gland diseases using the nano optical sensor thin film Sm-doxycycline complex.
Durable diagnosis of seminal vesicle and sexual gland diseases using the nano optical sensor thin film Sm-doxycycline complex.
Analytica chimica acta (2014-06-24)
M S Attia, A O Youssef, R H El-Sherif
ABSTRACT
A new method in which a nano optical sensor for diagnosis of different diseases of seminal vesicle and sexual gland was prepared. The working principle of the method depends on the determination of the fructose concentration in semen of different patients by using nano optical sensor thin film Sm-doxycycline doped in sol-gel matrix. The assay is based on the quenching of the characteristic emission bands of Sm(3+) present in silica doped Sm-doxycycline nanooptode thin film by different fructose concentrations in acetonitrile at λex = 400 nm. This method was optimized for parameters, such as, solvent effect, operational stability, shelf life and interference parameters. Good and reproducible linearity (1 × 10(-9) - 5.0 × 10(-5) mol L(-1)) with a detection limit of 9.0 × 10(-10) mol L(-1) and quantification limit of detection (LOQ) 2.7 × 10(-9) mol L(-1) were obtained. Seminal fructose determination in different patient samples after appropriate dilutions confirmed the reliability of this technique. The method was successfully applied for routine fructose monitoring in human semen samples of different cases such as; obstructive and non-obstructive azoospermia, inflammation of male accessory glands, atrophy of seminal vesicle, congenital vas deferens and retrograde ejaculation.
Sigma-Aldrich
Diethoxydimethylsilane, 97%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Diethoxydimethylsilane, purum, ≥97.0% (GC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material