Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
131903
Tetraethyl orthosilicate
reagent grade, 98%
Synonym(s):
Orthosilicic acid tetraethyl ester, Silicon tetraethoxide, TEOS, Tetraethoxysilane, Tetraethoxysilicon(IV), Tetraethyl silicate
About This Item
Recommended Products
grade
reagent grade
Quality Level
vapor density
7.2 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
Assay
98%
form
liquid
refractive index
n20/D 1.382 (lit.)
bp
168 °C (lit.)
density
0.933 g/mL at 20 °C (lit.)
SMILES string
CCO[Si](OCC)(OCC)OCC
InChI
1S/C8H20O4Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-8H2,1-4H3
InChI key
BOTDANWDWHJENH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- As an anti-solvent to fabricate efficient perovskite layer for inverted planar solar cells with enhanced power conversion efficiency and reproducibility. TEOS helps to achieve high-quality perovskite films with reduced density of defects, and thus slower carrier recombination.
- As a sol-gel precursor to synthesize SiO2-TiO2 anti-reflective self-cleaning coatings for solar cells.
- To fabricate nano silica functionalized crosslinked hybrid membranes for proton exchange membrane fuel cell applications. These membranes have better proton conductivity and thermal stability.
- To prepare poly(vinylidene fluoride)/TEOS matrix separator to enhance the performance of Li-ion batteries. The separator exhibits excellent electrolyte retention and wettability.
Features and Benefits
- Excellent adhesion on substrates
- Uniform film deposition
- Cross-linker to introduce polymer strands into hydrogel
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
113.0 °F - closed cup
Flash Point(C)
45 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
Is anything else needed to use Product 131903, Tetraethyl Orthosilicate in a hydrolysis reaction?
1 answer-
An acid or alkali catalyst is needed to do a hydrolysis reaction to free the silanol group for use.
Helpful?
-
-
What is the freezing point of Product 131903, Tetraethyl Orthosilicate?
1 answer-
The freezing point is ~85°C.
Helpful?
-
-
How can I purify Product 131903, Tetraethyl Orthosilicate?
1 answer-
Fractionate through an 80cm Podbielniak type column with heated jacket and partial take-off head. [Purification of Laboratory Chemicals Fourth Ed]
Helpful?
-
-
Product 131903, Tetraethyl orthosilicate is commonly used as a precursor to prepare xerogel. What is xerogel?
1 answer-
A xerogel is a solid formed from a gel by drying with unhindered shrinkage. Xerogel usually retains high porosity and large surfaced area, along with very small pore size.
Helpful?
-
-
What is the solubility of Product 131903, Tetraethyl Orthosilicate?
1 answer-
According to the 13th Edition of the chemicals encyclopedia published by the Royal Society of Chemistry, Tetraethyl Orthosilicate is practically insoluble in water and is slowly decomposed by it. It is miscible with alcohol.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service