Merck
  • Raphe AMPA receptors and nicotinic acetylcholine receptors mediate ketamine-induced serotonin release in the rat prefrontal cortex.
Raphe AMPA receptors and nicotinic acetylcholine receptors mediate ketamine-induced serotonin release in the rat prefrontal cortex.
The international journal of neuropsychopharmacology (2014-05-24)
Naoya Nishitani, Kazuki Nagayasu, Nozomi Asaoka, Mayumi Yamashiro, Hisashi Shirakawa, Takayuki Nakagawa, Shuji Kaneko
ABSTRACT
Several lines of evidence indicate that ketamine has a rapid antidepressant-like effect in rodents and humans, but underlying mechanisms are unclear. In the present study, we investigated the effect of ketamine on serotonin (5-HT) release in the rat prefrontal cortex by in vivo microdialysis. A subcutaneous administration of ketamine (5 and 25 mg/kg) significantly increased the prefrontal 5-HT level in a dose-dependent manner, which was attenuated by local injection of α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR) antagonists into the dorsal raphe nucleus (DRN). Direct stimulation of AMPARs in the DRN significantly increased prefrontal 5-HT level, while intra-DRN injection of ketamine (36.5 nmol) had no effect. Furthermore, intra-DRN injection of an α 4 β 2-nicotinic acetylcholine receptor (nAChR) antagonist, dihydro-β-erythroidine (10 nmol), significantly attenuated the subcutaneous ketamine-induced increase in prefrontal 5-HT levels. These results suggest that AMPARs and α 4 β 2-nAChRs in the DRN play a key role in the ketamine-induced 5-HT release in the prefrontal cortex.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, for ion-selective electrodes, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%