Merck
  • Repetitive injection method: a tool for investigation of injection zone formation and its compression in microfluidic liquid chromatography.
Repetitive injection method: a tool for investigation of injection zone formation and its compression in microfluidic liquid chromatography.
Journal of chromatography. A (2015-01-22)
Martin Gilar, Thomas S McDonald, Gregory Roman, Jay S Johnson, James P Murphy, James W Jorgenson
ABSTRACT
Sample introduction in microfluidic liquid chromatography often generates wide zones rather than peaks, especially when a large sample volume (relative to column volume) is injected. Formation of wide injection zones can be further amplified when the sample is dissolved in a strong eluent. In some cases sample breakthrough may occur, especially when the injection is performed into short trapping columns. To investigate the band formation and subsequent zone focusing under gradient elution in situations such as these, we developed the Repetitive Injection Method (RIM), based on the temporally resolved introduction of two discrete peaks to a column, mimicking both the leading and trailing edges of a larger, singly injected sample zone. Using titanium microfluidic 0.32 mm I.D. columns, the results of RIM experiments were practically identical to injection of a correspondingly larger single zone volume. It was also experimentally shown that zone width (spacing between two injected peaks) decreases during gradient elution. We utilized RIM experiments to investigate wide sample zones created by strong sample solvent, and subsequent gradient zone focusing for a series of compounds. This experimental work was compared with computationally simulated chromatograms. The success of sample focusing during injection and gradient elution depends not only on an analyte's absolute retention, but also on how rapidly the analyte's retention changes during the mobile phase gradient.
Product Description
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Valerophenone, 99%
Sigma-Aldrich
Hexanophenone, 99%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Indole, ≥99%
Supelco
Propiophenone, analytical standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetophenone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetophenone, natural, 98%, FG
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Butyrophenone, ≥99%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Reserpine, crystallized, ≥99.0% (HPLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Reserpine
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Uracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ultrapure Acetonitrile
Reserpine, European Pharmacopoeia (EP) Reference Standard