Skip to Content
Merck
All Photos(6)

Documents

U0750

Sigma-Aldrich

Uracil

≥99.0%

Synonym(s):

2,4(1H,3H)-Pyrimidinedione, 2,4-Dihydroxypyrimidine, 2,4-Pyrimidinediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O2
CAS Number:
Molecular Weight:
112.09
Beilstein:
507828
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥99.0%

form

powder

mp

>300 °C (lit.)

SMILES string

O=C1NC=CC(=O)N1

InChI

1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)

InChI key

ISAKRJDGNUQOIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Uracil belongs to the pyrimidine nucleobase family. It is naturally occurring and is an important and active part of RNA.

Application

Uracil has been used as:
  • a nucleotide standard to determine DNA methylation by high-performance liquid chromatography
  • a component of synthetic define medium for culturing Saccharomyces cerevisiae and its strain
  • a component of the amino acid supplement to culture Bacillus sp

Biochem/physiol Actions

Uracil and its derivatives play an integral part in medicinal chemistry for the synthesis of cancer, viral infections, autosomal recessive disorder, thyroid, and diabetic drugs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative metabolite profiling of wild type and thermo-tolerant mutant of Saccharomyces cerevisiae
Kim S, et al.
Process Biochemistry (Oxford, United Kingdom), 111, 62-68 (2021)
Deepthi Ramesh et al.
European journal of medicinal chemistry, 207, 112801-112801 (2020-09-15)
Uracil is one of the most notable pharmacophores in medicinal chemistry as the pyrimidine nucleobase forms an integral part of many commercial drugs. Though the name uracil is usually associated with cancer drugs, there are many uracil-based compounds which can
Sandra M Carvalho et al.
PloS one, 8(3), e58492-e58492 (2013-03-19)
Links between carbohydrate metabolism and virulence in Streptococcus pneumoniae have been recurrently established. To investigate these links further we developed a chemically defined medium (CDM) and standardized growth conditions that allowed for high growth yields of the related pneumococcal strains
Daisuke Katagiri et al.
Journal of the American Society of Nephrology : JASN, 24(12), 2034-2043 (2013-10-05)
Accumulating evidence of the beyond-glucose lowering effects of a gut-released hormone, glucagon-like peptide-1 (GLP-1), has been reported in the context of remote organ connections of the cardiovascular system. Specifically, GLP-1 appears to prevent apoptosis, and inhibition of dipeptidyl peptidase-4 (DPP-4)
Pietro Andreone et al.
Gastroenterology, 147(2), 359-365 (2014-05-14)
The interferon-free regimen of ABT-450 (a protease inhibitor), ritonavir, ombitasvir (an NS5A inhibitor), dasabuvir (a non-nucleoside polymerase inhibitor), and ribavirin has shown efficacy in patients with hepatitis C virus (HCV) genotype 1b infection-the most prevalent subgenotype worldwide. We evaluated whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service