Merck
  • Elafin drives poor outcome in high-grade serous ovarian cancers and basal-like breast tumors.
Elafin drives poor outcome in high-grade serous ovarian cancers and basal-like breast tumors.
Oncogene (2014-01-29)
S I Labidi-Galy, A Clauss, V Ng, S Duraisamy, K M Elias, H-Y Piao, E Bilal, R A Davidowitz, Y Lu, G Badalian-Very, B Györffy, U-B Kang, S Ficarro, S Ganesan, G B Mills, J A Marto, R Drapkin
ABSTRACT
High-grade serous ovarian carcinoma (HGSOC) and basal-like breast cancer (BLBC) share many features including TP53 mutations, genomic instability and poor prognosis. We recently reported that Elafin is overexpressed by HGSOC and is associated with poor overall survival. Here, we confirm that Elafin overexpression is associated with shorter survival in 1000 HGSOC patients. Elafin confers a proliferative advantage to tumor cells through the activation of the MAP kinase pathway. This mitogenic effect can be neutralized by RNA interference, specific antibodies and a MEK inhibitor. Elafin expression in patient-derived samples was also associated with chemoresistance and strongly correlates with bcl-xL expression. We extended these findings into the examination of 1100 primary breast tumors and six breast cancer cell lines. We observed that Elafin is overexpressed and secreted specifically by BLBC tumors and cell lines, leading to a similar mitogenic effect through activation of the MAP kinase pathway. Here too, Elafin overexpression is associated with poor overall survival, suggesting that it may serve as a biomarker and therapeutic target in this setting.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
L-Threonine
Serine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Human Trappin-2 / Elafin  ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Tyrosine, 99%
Supelco
2-Butanone, analytical standard
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)