Skip to Content
Merck
All Photos(1)

Key Documents

T1340000

L-Threonine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(2S,3R)-2-Amino-3-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721646
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference standard

API family

l-threonine

manufacturer/tradename

EDQM

mp

256 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](O)[C@H](N)C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1

InChI key

AYFVYJQAPQTCCC-GBXIJSLDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

ʟ-Threonine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xunyan Dong et al.
Biotechnology advances, 29(1), 11-23 (2010-08-07)
L-threonine is an essential amino acid for mammals and as such has a wide and expanding application in industry with a fast growing market demand. The major method of production of l-threonine is microbial fermentation. To optimize L-threonine production the
X Li et al.
Journal of internal medicine, 275(4), 418-427 (2013-11-19)
Biochemical changes in the cerebrospinal fluid (CSF) could reflect pathophysiological processes in Alzheimer's disease (AD). However, it is still not clear how these processes correlate with grey matter (GM) volume and microstructural changes in the brain. To assess the relationship
Synthetic N- and O-glycosyl derivatives of L-asparagine, L-serine, and L-threonine.
H G Garg et al.
Advances in carbohydrate chemistry and biochemistry, 43, 135-201 (1985-01-01)
Yi-Fang Chang et al.
Oncotarget, 6(1), 85-100 (2014-11-22)
Angiogenesis, one of the major routes for tumor invasion and metastasis represents a rational target for therapeutic intervention. Recent development in drug discovery has highlighted the diverse biological and pharmacological properties of hydroxamate derivatives. In this study, we characterized the
Na Wang et al.
Cancer, 120(19), 2965-2979 (2014-06-06)
The telomerase reverse transcriptase (TERT) promoter mutations C228T and C250T have been found in many malignancies, including in thyroid carcinomas. However, it is unclear how early these mutations occur in thyroid tumorigenesis. The study included primary tumors from 58 patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service