Skip to Content
Merck
All Photos(3)

Key Documents

360473

Sigma-Aldrich

2-Butanone

ACS reagent, ≥99.0%

Synonym(s):

Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
741880
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
80 °C (lit.)
vapor pressure:
71 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

2.49 (vs air)

vapor pressure

71 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

960 °F

expl. lim.

10.1 %

dilution

(for analytical testing)

impurities

≤0.0005 meq/g Titr. acid
≤0.20% water

evapn. residue

≤0.0025%

color

APHA: ≤15

refractive index

n20/D 1.379 (lit.)

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Butanone is an organic compound that can be used in the chemical industry as an oxidative reaction activator, extractive agent, solvent, and chemical intermediate. It can be produced by the catalytic dehydrogenation of 2-butanol.

Application

2-Butanonecan be used as a solvent:
  • To synthesize waterborne polyurethanes by reacting polyols and diisocyanates via polymerization reaction.
  • In the Al-catalyzed chemoselective coupling of oxetanes and carbon dioxide to synthesize six-membered cyclic carbonates.

Features and Benefits

  • Low-boiling solvent
  • High power of dissolution
  • High ratio of dissolved matter to viscosity
  • Miscible with a wide range of hydrocarbonswithout affecting the solid′s content or viscosity
  • Favorable volume/mass ratio due to its low density
  • Fast evaporation rate

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A volumetric and viscosity study for the mixtures of 1-n-butyl-3-methylimidazolium tetrafluoroborate ionic liquid with acetonitrile, dichloromethane, 2-butanone and N, N-dimethylformamide.
Wang J, et al.
Green Chemistry, 5(5), 618-622 (2003)
A simple one-pot multicomponent synthesis of an octahedral nanocontainer molecule.
Liu X and Warmuth R.
Nature Protocols, 2(5), 1288-1296 (2007)
C Philippe et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 69(9), 1212-1217 (2011-05-10)
[(11)C]PIB is still the standard PET compound for Alzheimer imaging targeting beta-amyloid plaques. We aimed to establish a fully-automated procedure for the synthesis and purification of [(11)C]PIB with a high degree of reliability and improved specific activity as well as
L B Goldsmith et al.
Contact dermatitis, 19(5), 348-350 (1988-11-01)
Irritant contact dermatitis along with an increased transepidermal water loss can result from exposing the skin to solvents. A study of the interaction of various solvents with human stratum corneum was made using thin-layer chromatography. Comparison of 10 solvents (trichloroethylene
Consalvo Petti et al.
Oncotarget, 6(1), 221-233 (2014-12-05)
Constitutively active receptor tyrosine kinases (RTKs) are known oncogenic drivers and provide valuable therapeutic targets in many cancer types. However, clinical efficacy of RTK inhibitors is limited by intrinsic and acquired resistance. To identify genes conferring resistance to inhibition of

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service