145726
DL-Tyrosine
99%
Synonym(s):
(±)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-DL-alanine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
4-(HO)C6H4CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
181.19
Beilstein:
515881
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
mp
≥300 °C (lit.)
application(s)
peptide synthesis
SMILES string
NC(Cc1ccc(O)cc1)C(O)=O
InChI
1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)
InChI key
OUYCCCASQSFEME-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
W D Phillips et al.
Experimental neurology, 270, 29-40 (2014-12-30)
Muscle-specific tyrosine kinase (MuSK) autoantibodies are the hallmark of a form of myasthenia gravis (MG) that can challenge the neurologist and the experimentalist. The clinical disease can be difficult to treat effectively. MuSK autoantibodies affect the neuromuscular junction in several
Philippe Menasché et al.
European heart journal, 36(12), 743-750 (2014-05-20)
There is now compelling evidence that cells committed to a cardiac lineage are most effective for improving the function of infarcted hearts. This has been confirmed by our pre-clinical studies entailing transplantation of human embryonic stem cell (hESC)-derived cardiac progenitors
Shannon L Maude et al.
Blood, 125(11), 1759-1767 (2015-02-04)
Early T-cell precursor (ETP) acute lymphoblastic leukemia (ALL) is a recently described subtype of T-ALL characterized by a unique immunophenotype and genomic profile, as well as a high rate of induction failure. Frequent mutations in cytokine receptor and Janus kinase
Dana Shani Galili et al.
Current biology : CB, 24(15), 1712-1722 (2014-07-22)
Drosophila learn to avoid odors that are paired with aversive stimuli. Electric shock is a potent aversive stimulus that acts via dopamine neurons to elicit avoidance of the associated odor. While dopamine signaling has been demonstrated to mediate olfactory electric
Selective neuronal differentiation of neural stem cells induced by nanosecond microplasma agitation.
Z Xiong et al.
Stem cell research, 12(2), 387-399 (2014-01-01)
An essential step for therapeutic and research applications of stem cells is their ability to differentiate into specific cell types. Neuronal cells are of great interest for medical treatment of neurodegenerative diseases and traumatic injuries of central nervous system (CNS)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service