Merck
  • Microbial production of aliphatic (S)-epoxyalkanes by using Rhodococcus sp. strain ST-10 styrene monooxygenase expressed in organic-solvent-tolerant Kocuria rhizophila DC2201.
Microbial production of aliphatic (S)-epoxyalkanes by using Rhodococcus sp. strain ST-10 styrene monooxygenase expressed in organic-solvent-tolerant Kocuria rhizophila DC2201.
Applied and environmental microbiology (2015-01-04)
Hiroshi Toda, Takuya Ohuchi, Ryouta Imae, Nobuya Itoh
ABSTRACT
We describe the development of biocatalysis for producing optically pure straight-chain (S)-epoxyalkanes using styrene monooxygenase of Rhodococcus sp. strain ST-10 (RhSMO). RhSMO was expressed in the organic solvent-tolerant microorganism Kocuria rhizophila DC2201, and the bioconversion reaction was performed in an organic solvent-water biphasic reaction system. The biocatalytic process enantioselectively converted linear terminal alkenes to their corresponding (S)-epoxyalkanes using glucose and molecular oxygen. When 1-heptene and 6-chloro-1-hexene were used as substrates (400 mM) under optimized conditions, 88.3 mM (S)-1,2-epoxyheptane and 246.5 mM (S)-1,2-epoxy-6-chlorohexane, respectively, accumulated in the organic phase with good enantiomeric excess (ee; 84.2 and 95.5%). The biocatalysis showed broad substrate specificity toward various aliphatic alkenes, including functionalized and unfunctionalized alkenes, with good to excellent ee. Here, we demonstrate that this biocatalytic system is environmentally friendly and useful for producing various enantiopure (S)-epoxyalkanes.
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Supelco
Styrene, analytical standard
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Supelco
Hexadecane, analytical standard
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Supelco
Styrene, analytical standard
Sigma-Aldrich
6-Chloro-1-hexene, 96%
Sigma-Aldrich
1-Heptene, 97%
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
2-Methyl-1-hexene, 96%
Sigma-Aldrich
1-Heptene, ≥99%
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dodecane, analytical standard
Supelco
Density Standard 749 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Di-n-octyl phthalate, analytical standard
Sigma-Aldrich
Tetradecane, olefin free, ≥99.0% (GC)
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Sigma-Aldrich
Tetradecane, ≥99%
Supelco
Tetradecane, analytical standard
Sigma-Aldrich
6-Bromo-1-hexene, 95%
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Diethyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Di-n-octyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland