Skip to Content
Merck
All Photos(3)

Key Documents

296317

Sigma-Aldrich

Hexadecane

anhydrous, ≥99%

Synonym(s):

n-Hexadecane, Cetane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CH3
CAS Number:
Molecular Weight:
226.44
Beilstein:
1736592
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
287 °C (lit.)
vapor pressure:
1 mmHg ( 105.3 °C)

grade

anhydrous

Quality Level

vapor density

7.8 (vs air)

vapor pressure

1 mmHg ( 105.3 °C)

Assay

≥99%

form

liquid

autoignition temp.

395 °F

impurities

<0.003% water
<0.005% water (100 mL pkg)

refractive index

n20/D 1.434 (lit.)

bp

287 °C (lit.)

mp

18 °C (lit.)

transition temp

solidification point 17.5-18.5 °C

density

0.773 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexadecane may be used as a solvent for trioctylamine-based synthesis of indium antimonide (InSb) nanocrystals. It may also be used as an internal standard for the synthesis of poly(9,9-
dioctylfluorene)-b-poly(3-hexylthiophene) via Grignard metathesis chain growth polymerization.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hajime Shinoda et al.
Communications biology, 4(1), 476-476 (2021-04-21)
CRISPR-based nucleic-acid detection is an emerging technology for molecular diagnostics. However, these methods generally require several hours and could cause amplification errors, due to the pre-amplification of target nucleic acids to enhance the detection sensitivity. Here, we developed a platform
Chain-growth synthesis of polyfluorenes with low polydispersities, block copolymers of fluorene, and end-capped polyfluorenes: toward new optoelectronic materials
Javier AE, et al.
Macromolecules, 43(7), 3233-3237 (2010)
Maartje L J Steegmans et al.
Langmuir : the ACS journal of surfaces and colloids, 25(17), 9751-9758 (2009-07-09)
Emulsification in microdevices (microfluidic emulsification) involves micrometer-sized droplets and fast interface expansion rates. In addition, droplets are formed in less than milliseconds, and therefore traditional tensiometric techniques cannot be used to quantify the actual interfacial tension. In this paper, monodisperse
Excess volumes for n-alkanols+ n-alkanes IV. Binary mixtures of decan-1-ol+ n-pentane,+ n-hexane,+ n-octane,+ n-decane, and+ n-hexadecane.
Treszczanowicz AJ, et al.
The Journal of Chemical Thermodynamics, 13(3), 253-260 (1981)
Colloidal synthesis of InSb nanocrystals with controlled polymorphism using indium and antimony amides.
Yarema M and Kovalenko MV
Chemistry of Materials, 25(9), 1788-1792 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service