Skip to Content
Merck
All Photos(1)

Documents

44010

Supelco

Dodecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CH3
CAS Number:
Molecular Weight:
170.33
Beilstein:
1697175
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.96 (vs air)

vapor pressure

1 mmHg ( 47.8 °C)

Assay

≥99.8% (GC)

autoignition temp.

401 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.421 (lit.)
n20/D 1.421

bp

215-217 °C (lit.)

mp

−9.6 °C (lit.)

density

0.75 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCC

InChI

1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3

InChI key

SNRUBQQJIBEYMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dodecane is a clear, colorless liquid, having a mild aliphatic hydrocarbon odor, and belongs to the family of acyclic alkanes.

Application

The analytical standard can also be used as follows:

  • Simultaneous determination of volatile organic compounds by gas chromatography
  • Determination of seven volatile organic compounds in the surgically resected colonic tissue samples of colorectal cancer patients by gas chromatography-mass spectrometry (GC-MS) following their extraction by headspace- and direct injection- based solid phase microextraction (SPME)
  • Measurement of concentrations of volatile organic compounds (VOCs) in biogas samples obtained from three different biogas plants by gas chromatography-mass spectrometry
  • Quantification of 29 organic air pollutants, consisting of alkanes, carbonyls, terpenes, and aromatic hydrocarbons, in the indoor air of 22 houses using passive samplers
  • Gas chromatography-mass spectrometry (GC-MS) method-based profiling of volatile organic compounds in the olive fruit headspace extracts

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F - Pensky-Martens closed cup

Flash Point(C)

70 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Enzyme-catalysed siloxane bond formation
Bassindale RA, et al.
Journal of Inorganic Biochemistry, 96(2-3), 401-406 (2003)
Groundwater Chemicals Desk Reference, Fourth Edition (2007)
Label?Free Biomedical Imaging of Lipids by Stimulated Raman Scattering Microscopy
Ramachandran VP, et al.
Current Protocols in Molecular Biology, 109(1), 30-33 (2015)
Hong Sun et al.
Aging, 12(20), 20432-20444 (2020-10-29)
In human macrophages and monocytes, lipopolysaccharide (LPS) induces nuclear factor kappa B (NFκB) activation and pro-inflammatory cytokines production. We tested the possible involvement of Ku70 and Ku80 in the process. In THP-1 macrophages and primary human peripheral blood mononuclear cells
X-L Cao et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 28(6), 791-798 (2011-05-31)
A total of 154 food composite samples from the 2008 total diet study in Quebec City were analysed for bisphenol A (BPA), and BPA was detected in less than half (36%, or 55 samples) of the samples tested. High concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service