Merck
  • Simultaneous determination of sulfonamides and metabolites in manure samples by one-step ultrasound/microwave-assisted solid-liquid-solid dispersive extraction and liquid chromatography-mass spectrometry.
Simultaneous determination of sulfonamides and metabolites in manure samples by one-step ultrasound/microwave-assisted solid-liquid-solid dispersive extraction and liquid chromatography-mass spectrometry.
Analytical and bioanalytical chemistry (2015-03-04)
Hui-zhen Wu, Ming-rong Qian, Jian-mei Wang, Hu Zhang, Jun-wei Ma, Zu-guang Li, Maw-rong Lee
ABSTRACT
An in-line matrix cleanup method was used for the simultaneous extraction of 15 sulfonamides and two metabolites from manure samples. The ultrasound/microwave-assisted extraction (UMAE) combined with solid-liquid-solid dispersive extraction (SLSDE) procedure provides a simple sample preparation approach for the processing of manure samples, in which the extraction and cleanup are integrated into one step. Ultrasonic irradiation power, extraction temperature, extraction time, and extraction solvent, which could influence the UMAE efficiency, were investigated. C18 was used as the adsorbent to reduce the effects of interfering components during the extraction procedure. The extracts were concentrated, and the analytes were analyzed by liquid chromatography-tandem mass spectrometry (LC-MS/MS) without any further cleanup. The isotopically labeled compounds sulfamethoxazole-d 4, sulfamethazine-d 4, sulfamonomethoxine-d 4, and sulfadimethoxine-d 6 were selected as internal standards to minimize the matrix effect in this method. The recoveries of the antibiotics tested ranged from 71 to 118 % at the three spiking levels examined (20, 200, and 500 μg · kg(-1)). The limits of detections were 1.2-3.6 μg · kg(-1) and the limits of quantification were 4.0-12.3 μg · kg(-1) for the sulfonamides and their metabolites. The applicability of the method was demonstrated by analyzing 30 commercial manure samples. The results indicated that UMAE-SLSDE combined with LC-MS/MS is a simple, rapid, and environmentally friendly method for the analysis of sulfonamides and their metabolites in manure, and it could provide the basis for a risk assessment of the antibiotics in agricultural environments.
Product Description
Supelco
Sulfamethoxypyridazine, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfamonomethoxine
Sigma-Aldrich
Sulfameter
Supelco
Sulfadoxin, VETRANAL®, analytical standard
Supelco
Sulfapyridine melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfamonomethoxine, VETRANAL®, analytical standard
Sulfadoxine, European Pharmacopoeia (EP) Reference Standard
Sulfafurazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfamethoxypyridazine, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Sulfapyridine, ≥99.0%
Sigma-Aldrich
Sulfamerazine, ReagentPlus®, ≥99.0%
Supelco
Acetonitrile, analytical standard