Skip to Content
Merck
All Photos(1)

Key Documents

46858

Supelco

Sulfamethoxypyridazine

VETRANAL®, analytical standard

Synonym(s):

N1-(6-Methoxypyridazin-3-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O3S
CAS Number:
Molecular Weight:
280.30
Beilstein:
277076
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(NS(=O)(=O)c2ccc(N)cc2)nn1

InChI

1S/C11H12N4O3S/c1-18-11-7-6-10(13-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,15)

InChI key

VLYWMPOKSSWJAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfamethoxypyridazine is a sulfonamide antibiotic mainly used to prevent infections as well as conditions such as coccidiosis, diarrhea and gastroenteritis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfamethoxypyridazine may be used as a reference standard for the determination of sulfamethoxypyridazine in pharmaceutical formulations by liquid chromatography method.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Bustamante et al.
The Journal of pharmacy and pharmacology, 47(7), 550-555 (1995-07-01)
The solubility of sulphamethoxypyridazine was measured at several temperatures in mixtures of water:ethanol and ethanol:ethyl acetate. Sulphamethoxypyridazine was chosen as a model drug to compare the solvation effects of proton donor-proton acceptor (water and ethanol) and proton acceptor (ethyl acetate)
S K Garg et al.
Veterinary research, 28(1), 101-104 (1997-01-01)
The pharmacokinetics of sulfamethoxypyridazine (SMP) was investigated in goats after a single intramuscular (im) or subcutaneous (sc) administration (100 mg/kg body weight). The biological half lives of SMP following im and sc administration were found to be 11.0 and 13.7
D Bonazzi et al.
Farmaco (Societa chimica italiana : 1989), 49(6), 381-386 (1994-06-01)
Derivative and difference spectrophotometric methods are described for the direct simultaneous analysis of combinations of Trimethoprim with sulfonamide drugs (sulfadiazine, sulfamethoxazole, sulfamethoxypyridazine) in commercial dosage forms. The more advantageous approach (derivative and difference mode) is suggested for each binary mixture.
M Thornhill et al.
The British journal of dermatology, 143(1), 117-126 (2000-07-25)
Twenty-five patients with mucous membrane pemphigoid (MMP), whose oral lesions were unresponsive to topical steroid treatment, were treated with 1 g daily of sulphamethoxypyridazine (SMXP), a long-acting sulphonamide antibiotic, in an open prospective clinical trial. Lesion severity was assessed objectively
W T Hughes et al.
Antimicrobial agents and chemotherapy, 40(4), 962-965 (1996-04-01)
A remarkably high rate of adverse events is associated with the use of trimethoprim-sulfamethoxazole in patients with human immunodeficiency virus type 1 infection. We examined the efficacies of sulfonamides alone in the prevention of Pneumocystis carinii pneumonitis, with the assumption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service