Merck
  • Trimethylamine-N-oxide: a carnitine-derived metabolite that prolongs the hypertensive effect of angiotensin II in rats.
Trimethylamine-N-oxide: a carnitine-derived metabolite that prolongs the hypertensive effect of angiotensin II in rats.
The Canadian journal of cardiology (2014-12-06)
Marcin Ufnal, Radoslaw Jazwiec, Michal Dadlez, Adrian Drapala, Mariusz Sikora, Janusz Skrzypecki
ABSTRACT
Recent evidence suggests that an elevated plasma trimethylamine N-oxide (TMAO) level is associated with an increased risk of adverse cardiovascular events in humans; however, the mechanism is not clear. The aims of this study were to establish the plasma TMAO level in rats and to evaluate the effect of TMAO on arterial blood pressure (BP) and the hemodynamic effects of angiotensin II (Ang II). Twelve-week-old, Sprague-Dawley rats were implanted with telemetric transmitters, and continuous recordings of heart rate, systolic BP (SBP), and diastolic BP (DBP) were made for 7 days before and 14 days during osmotic minipump-driven subcutaneous infusion of saline (controls), TMAO, low-dose Ang II, or Ang II + TMAO. Plasma TMAO concentration was evaluated using liquid chromatography coupled with triple-quadrupole mass spectrometry. The plasma TMAO concentration in controls was 0.57 μmol/L, whereas in TMAO-infused rats it was 58 μmol/L. Neither saline nor TMAO infusion affected SBP and DBP. Infusion of Ang II significantly increased SBP and DBP for the first 5 days of infusion only. In contrast, infusion of Ang II + TMAO produced a hypertensive response that lasted until the end of the experiment. TMAO infusions did not affect body weight and motor activity. We showed that physiological plasma TMAO concentration in rats was approximately 10 times lower than that reported in humans. Furthermore, the new finding of the study is that TMAO does not affect BP in normotensive animals. However, it prolongs the hypertensive effect of Ang II.
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trimethylamine-13C3,15N hydrochloride, 99 atom % 13C, 98 atom % 15N
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Trimethylamine N-oxide, 95%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard